Skip to Content
Merck
All Photos(1)

Key Documents

F5506

Sigma-Aldrich

Freund′s Adjuvant, Incomplete

liquid

Synonym(s):

FICA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

form

liquid

Quality Level

color

faintly yellow to yellow

storage temp.

2-8°C

Application

Freund′s Adjuvants may be used to produce water-in-oil emulsions of immunogens. Antigens in water-in-oil emulsions stimulate high and long-lasting antibody responses which can be attributed to the slow release of antigen.
Freund′s adjuvant (incomplete) lacks the Mycobacteria found in Complete Freund′s Adjuvant so it minimizes the side-effects.Freund′s Adjuvant, Incomplete is used for boost injections. It was used to emulsify antigens that are injected into animals to produce monoclonal antibodies from the antiserum.[1][2]

Other Notes

Each mL contains 0.85 mL paraffin oil and 0.15 mL mannide monooleate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Gillett et al.
PloS one, 4(11), e7773-e7773 (2009-11-17)
Multiple Sclerosis (MS) is a chronic inflammatory disease causing demyelination and nerve loss in the central nervous system. Experimental autoimmune encephalomyelitis (EAE) is an animal model of MS that is widely used to investigate complex pathogenic mechanisms. Transcriptional control through
Elisabeth J M Huijbers et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(11), 4535-4544 (2010-07-17)
Monoclonal antibody-based therapies have made an important contribution to current treatment strategies for cancer and autoimmune disease. However, the cost for these new drugs puts a significant strain on the health-care economy, resulting in limited availability for patients. Therapeutic vaccination
Stephan Culemann et al.
Nature, 572(7771), 670-675 (2019-08-09)
Macrophages are considered to contribute to chronic inflammatory diseases such as rheumatoid arthritis1. However, both the exact origin and the role of macrophages in inflammatory joint disease remain unclear. Here we use fate-mapping approaches in conjunction with three-dimensional light-sheet fluorescence
J Ziebuhr et al.
Journal of virology, 69(7), 4331-4338 (1995-07-01)
The RNA polymerase gene of human coronavirus (HCV) 229E encodes a large polyprotein that contains domains with motifs characteristic of both papain-like cysteine proteinases and proteinases with homology to the 3C proteinase of picornaviruses. In this study, we have, first
Katrine Hygum et al.
PloS one, 6(5), e20638-e20638 (2011-06-10)
In humans, the cobalamin (Cbl) -binding protein transcobalamin (TC) transports Cbl from the intestine and into all the cells of the body, whereas the glycoprotein haptocorrin (HC), which is present in both blood and exocrine secretions, is able to bind

Articles

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Questions

  1. How long it is expired

    1 answer
    1. This product is not assigned an expiration date. It is assigned a recommended retest date of approximately 10 months after the quality release date. A recommended retest date is the period of time during which the product is expected to remain within established stability specifications, provided that it has been stored under defined conditions. After the Retest Date, product samples should be examined to ensure that the product is still in compliance with the established specifications. For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service