Skip to Content
Merck
All Photos(2)

Key Documents

F5387

Sigma-Aldrich

Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fc specific)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
indirect immunofluorescence: 1:32

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.

Specificity

Goat polyclonal anti-Mouse IgG (Fc specific)–FITC antibody detects all mouse IgG subclasses, it shows no reactivity with mouse IgG Fab fragment or human IgG. Identity and purity of the antibody is established by immunoelectrophoresis, prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion against anti-goat IgG and anti-goat whole serum result in single arcs of precipitation. Goat anti-Mouse IgG is conjugated to FITC and then purified by gel filtration to remove free FITC.

Immunogen

Purified mouse IgG Fc fragment

Application

Anti-Mouse IgG (Fc specific)-FITC antibody has be used for
  • Immunofluorescence
  • Immunohistochemistry
  • Immunostaining
  • Confocal microscopy
  • Flow cytometry

Biochem/physiol Actions

IgG antibody provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections IgG is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Anti-Mouse IgG (Fc specific)-FITC antibody is specific for Fc fragment of all mouse IgG subclasses. It shows no reactivity with mouse IgG Fab fragment or human IgG. Goat anti-Mouse IgG is conjugated to FITC and then purified by gel filtration to remove free FITC.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Boutinaud et al.
Journal of dairy science, 95(1), 177-187 (2011-12-24)
To assess the regulation of mammary cell activity, survival, and proliferation by prolactin (PRL), 5 Holstein cows in early lactation received daily i.m. injections of 1mg of quinagolide, a suppressor of PRL release, for 9 wk, whereas 4 control cows
César Díaz-Godínez et al.
Antioxidants (Basel, Switzerland), 10(6) (2021-07-03)
NETosis is a neutrophil process involving sequential steps from pathogen detection to the release of DNA harboring antimicrobial proteins, including the central generation of NADPH oxidase dependent or independent ROS. Previously, we reported that NETosis triggered by Entamoeba histolytica trophozoites
Muneer G Hasham et al.
Disease models & mechanisms, 10(3), 259-270 (2017-03-03)
Systemic autoimmune diseases such as systemic lupus erythematosus (SLE) and rheumatoid arthritis (RA) show significant heart involvement and cardiovascular morbidity, which can be due to systemically increased levels of inflammation or direct autoreactivity targeting cardiac tissue. Despite high clinical relevance
Shuxuan Li et al.
Viruses, 10(1) (2018-01-25)
Zika virus (ZIKV) has been defined as a teratogenic pathogen behind the increased number of cases of microcephaly in French Polynesia, Brazil, Puerto Rico, and other South American countries. Experimental studies using animal models have achieved tremendous insight into understanding
C Souriau et al.
Nucleic acids research, 25(8), 1585-1590 (1997-04-15)
Studies on receptor-ligand interactions are important for the design of agonists or antagonists of natural ligands. We developed a luciferase reporter assay to screen epidermal growth factor receptor (EGFR) binding molecules rapidly for their ability to stimulate or inhibit signal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service