Skip to Content
Merck

F5180

Anti-Fibroblast Growth Factor-Basic antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BFGF, Anti-FGF2, Anti-FGFB, Anti-HBGF-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, Neutral, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

bovine, human

technique(s)

immunohistochemistry: 5-15 μg/mL, neutralization: suitable, western blot: 0.1-0.2 μg/mL

UniProt accession no.

application(s)

research pathology

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... FGF2(281161)
human ... FGF2(2247)

Immunogen

purified bovine fibroblast growth factor basic peptide (146 aa).

Biochem/physiol Actions

Fibroblast Growth Factor Basic (also known as bFGF) is a pleotropic cytokine that that is a potent mitogenic factor. On binding to its cognate receptor tyrosine kinase, FGFR, bFGF is capable of activation of pathways such as MAPK and PI3K and produce cellular effects in varied cell types. Fibroblast Growth Factor-basic is reported to induce proliferation of fibroblasts, differentiation, wound healing and regeneration, limb formation, tissue remodeling and angiogenesis.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library