Skip to Content
Merck
All Photos(1)

Key Documents

F5126

Sigma-Aldrich

m-Fluoro-DL-phenylalanine

≥98%

Synonym(s):

3-Fluoro-DL-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
183.18
Beilstein:
2939807
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

m-Fluoro-DL-phenylalanine,

Assay

≥98%

Quality Level

form

powder

color

white to off-white

mp

240-250 °C

application(s)

cell analysis
peptide synthesis

SMILES string

NC(Cc1cccc(F)c1)C(O)=O

InChI

1S/C9H10FNO2/c10-7-3-1-2-6(4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)

InChI key

VWHRYODZTDMVSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

m-Fluoro-DL-phenylalanine, a toxic antimetabolite, is a racemic mixture of a substituted (halogenated) benzoyl D and L phenylalanine with potential use in antiviral and antimicrobial drug development.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica N Kinde et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(48), 13762-13767 (2016-11-20)
Voltage-gated sodium channels (NaV) play an important role in general anesthesia. Electrophysiology measurements suggest that volatile anesthetics such as isoflurane inhibit NaV by stabilizing the inactivated state or altering the inactivation kinetics. Recent computational studies suggested the existence of multiple
Nick J P Wierckx et al.
Applied and environmental microbiology, 71(12), 8221-8227 (2005-12-08)
Efficient bioconversion of glucose to phenol via the central metabolite tyrosine was achieved in the solvent-tolerant strain Pseudomonas putida S12. The tpl gene from Pantoea agglomerans, encoding tyrosine phenol lyase, was introduced into P. putida S12 to enable phenol production.
W D Kingsbury et al.
Journal of medicinal chemistry, 26(12), 1725-1729 (1983-12-01)
A series of di- and tripeptides containing D- and L-m-fluorophenylalanine was prepared and tested in vitro for the ability to inhibit the growth of the yeast Candida albicans. The results demonstrate that peptides containing L-m-fluorophenylalanine inhibited the growth of C.
H D Dettman et al.
Biophysical journal, 37(1), 243-251 (1982-01-01)
The nonlytic, filamentous coliphage M13 offers an excellent model system for the study of membrane-protein interactions. We prepare derivatives of the protein containing fluorine-labeled amino acids and use 19F nuclear magnetic resonance (NMR) to study the protein in both deoxycholate
Jennifer C Jackson et al.
Journal of the American Chemical Society, 129(5), 1160-1166 (2007-02-01)
19F NMR is a powerful tool for monitoring protein conformational changes and interactions; however, the inability to site-specifically introduce fluorine labels into proteins of biological interest severely limits its applicability. Using methods for genetically directing incorporation of unnatural amino acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service