Skip to Content
Merck
All Photos(2)

Key Documents

F4137

Sigma-Aldrich

Anti-Chicken IgY (IgG) (whole molecule)−FITC antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Chicken IgY (IgG) (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
indirect immunofluorescence: 1:200

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Chicken IgY is made of two heavy (H) and two light (L) chains. Its molecular weight is about ~180 kDa.
Chicken IgY is the major antibody found in chicken serum. Chickens generate a comparatively enhanced antibody response to mammalian antigenic proteins. These immunoglobulins do not bind to Fc receptors present in mammals and bacteria, and also do not interact with rheumatoid factors. Thus chicken antibodies are useful analytical tools for various research applications .

Specificity

Birds, reptiles and amphibians express IgY antibodies that are functionally equivalent to IgG. Anti-Chicken IgY (IgG) (whole molecule)-FITC antibody is specific for IgY of chicken. Rabbit Anti- Chicken IgY (IgG) is conjugated to fluorescein isothiocyanate (FITC). Free FITC is removed by gel filtration.

Immunogen

Chicken IgY (IgG)
Purified chicken IgY (IgG)

Application

Anti-Chicken IgY (IgG) (whole molecule) FITC antibody has been used:
  • in indirect immunofluorescence
  • in western blotting
  • in immunocytochemistry

Biochem/physiol Actions

IgY plays a major role in defense mechanism against systemic infections. It also possesses the ability to initiate anaphylactic reactions.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lineage conversion induced by pluripotency factors involves transient passage through an iPSC stage
Bar-Nur O, et al.
Nature Biotechnology, 33(7), 761-761 (2015)
Ori Bar-Nur et al.
Nature biotechnology, 33(7), 761-768 (2015-06-23)
Brief expression of pluripotency-associated factors such as Oct4, Klf4, Sox2 and c-Myc (OKSM), in combination with differentiation-inducing signals, has been reported to trigger transdifferentiation of fibroblasts into other cell types. Here we show that OKSM expression in mouse fibroblasts gives
Aymn T Abbas et al.
Vaccines, 8(4) (2020-11-04)
The Middle East respiratory syndrome coronavirus (MERS-CoV) was identified in 2012 and causes severe and often fatal acute respiratory illness in humans. No approved prophylactic and therapeutic interventions are currently available. In this study, we have developed egg yolk antibodies
Beatriz Maria de Almeida Braz et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 29(4), e014120-e014120 (2020-10-22)
Toxoplasmosis is a parasitic disease, which is prevalent across the world and has a strong environmental component in its transmission chain. The present study determined the seroprevalence and factors related to Toxoplasma gondii infection among free-range chickens from an Atlantic
Alexandre Dias Munhoz et al.
Veterinary parasitology, 206(3-4), 123-128 (2014-12-04)
The aim of this study was to experimentally evaluate infection in Gallus gallus domesticus with Neospora caninum tachyzoites of the NC-1 strain. Experimental infection was conducted in 90-day-old chickens, embryonated eggs and bioassays in dogs. In the first experiment, poults

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service