Skip to Content
Merck

Skip To

F3301

Factor Xa chromogenic substrate

chromogenic, ≥90% (HPLC), solid

Synonym(s):

CH3OCO-D-CHA-Gly-Arg-pNA-AcOH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Factor Xa chromogenic substrate, solid

assay

≥90% (HPLC)

Quality Segment

form

solid

mol wt

apparent mol wt 622.7

solubility

methanol: water (1:9): 25 mg/mL

storage temp.

2-8°C

General description

KM for human factor Xa is ~0.8mM

Application

Factor Xa chromogenic substrate has been used in factor Xa protease assay for placenta organelle protein and Prospirobolus joannsi.

Biochem/physiol Actions

Factor Xa chromogenic substrate contains peptide sequence Bz-Ile-Glu-Gly-Arg-pNA. Factor Xa mediates the release of p-NA chromogen post hydrolysis of substrate which is measurable photometrically.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3068H6660T2943
assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

solubility

methanol: water (1:9): 25 mg/mL

solubility

-

solubility

DMSO: soluble

solubility

methanol: water (1:9): 25 mg/mL

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

solid

form

powder

form

solid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

mol wt

apparent mol wt 622.7

mol wt

-

mol wt

2016

mol wt

apparent mol wt 658.9


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
F3301-25MG04061835556809

Questions

Reviews

No rating value

Active Filters