Skip to Content
Merck
All Photos(1)

Key Documents

F3272

Sigma-Aldrich

Inulin–FITC

from dahlia tuber

Synonym(s):

Fluorescein isothiocyanate–inulin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

dahlia tuber

Quality Level

form

powder

mol wt

2,000-5,000

color

light yellow to dark yellow, to dark orange

solubility

water: soluble, clear

storage temp.

2-8°C

Application


  • Development and validation of bioengineered intestinal tubules for translational research aimed at safety and efficacy testing of drugs and nutrients.: This study employs Inulin–FITC to investigate the absorption characteristics of bioengineered intestinal models, providing critical data on gut permeability and nutrient interaction, which is crucial for pharmaceutical and nutritional research (Jochems et al., 2019).

  • Role of Vitamin D in Maintaining Renal Epithelial Barrier Function in Uremic Conditions.: Utilizes Inulin–FITC to measure the efficacy of Vitamin D in preserving renal barrier function under stress conditions, which could lead to new treatments for chronic kidney disease (Mihajlovic et al., 2017).

  • Inhibition of p38 mitogen-activated protein kinase attenuates butyrate-induced intestinal barrier impairment in a Caco-2 cell monolayer model.: This investigation highlights the role of Inulin–FITC in studying the protective effects of kinase inhibition on intestinal barriers, relevant for gastrointestinal disorder treatments (Huang et al., 2014).

  • Measurement of glomerular filtration rate in conscious mice using FITC-inulin clearance.: Demonstrates the utility of Inulin–FITC in precise, non-invasive measurements of renal function, essential for nephrology research and therapy development (Qi and Breyer, 2009).

  • PPAR gamma agonist normalizes glomerular filtration rate, tissue levels of homocysteine, and attenuates endothelial-myocyte uncoupling in alloxan induced diabetic mice.: Incorporates Inulin–FITC to assess the efficacy of a PPAR gamma agonist in diabetic nephropathy, providing insights into metabolic disease management (Rodriguez et al., 2008).

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunhua Jin et al.
Hypertension (Dallas, Tex. : 1979), 53(6), 1048-1053 (2009-05-13)
Studies were designed to determine the effects of increases of renal perfusion pressure on the production of hydrogen peroxide (H(2)O(2)) and NO(2)(-)+NO(3)(-) within the renal outer medulla. Sprague-Dawley rats were studied with either the renal capsule intact or removed to
Paul Brakeman et al.
Biomicrofluidics, 10(6), 064106-064106 (2016-12-06)
Most current microfluidic cell culture systems are integrated single use devices. This can limit throughput and experimental design options, particularly for epithelial cells, which require significant time in culture to obtain a fully differentiated phenotype. In addition, epithelial cells require
Omar Cauli et al.
Neuromolecular medicine, 16(2), 360-375 (2013-12-18)
Treatment of patients with acute liver failure (ALF) is unsatisfactory and mortality remains unacceptably high. Blocking NMDA receptors delays or prevents death of rats with ALF. The underlying mechanisms remain unclear. Clarifying these mechanisms will help to design more efficient
Biman B Mandal et al.
Nanotechnology, 20(35), 355101-355101 (2009-08-13)
In recent times self-assembled micellar nanoparticles have been successfully employed in tissue engineering for targeted drug delivery applications. In this review, silk sericin protein from non-mulberry Antheraea mylitta tropical tasar silk cocoons was blended with pluronic F-127 and F-87 in
Richard B van Breemen et al.
Expert opinion on drug metabolism & toxicology, 1(2), 175-185 (2006-08-23)
Caco-2 cells are a human colon epithelial cancer cell line used as a model of human intestinal absorption of drugs and other compounds. When cultured as a monolayer, Caco-2 cells differentiate to form tight junctions between cells to serve as

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service