Skip to Content

F3023

α-1→(3,4)-Fucosidase solution from Xanthomonas sp.

buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32
EC Number:
232-914-2
MDL number:
EC Number:
Specific activity:
≥2 units/mg protein
Concentration:
≥0.5 unit/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

Quality Segment

specific activity

≥2 units/mg protein

packaging

vial of 0.02 unit

concentration

≥0.5 unit/mL

foreign activity

β-galactosidase, α-mannosidase, β-hexosaminidase, neuraminidase, α-1,2-fucosidase and proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Biochem/physiol Actions

Releases non-reducing, terminal α-1,3-fucose and α-1,4-fucose from complex carbohydrates.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 25 mM NaCl

Other Notes

One unit will release 1.0 μmole of fucose from Lewis X trisaccharide, 4-methylumbelliferyl glycoside per min at pH 5.0 at 37 °C.


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library