Skip to Content
Merck
All Photos(1)

Key Documents

F2802

Sigma-Aldrich

Fluvoxamine maleate

solid

Synonym(s):

(E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21F3N2O2 · C4H4O4
CAS Number:
Molecular Weight:
434.41
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

solubility

H2O: soluble

originator

Solvay

storage temp.

2-8°C

SMILES string

COCCCC/C(C1=CC=C(C(F)(F)F)C=C1)=N\OCCN.OC(/C=C\C(O)=O)=O

InChI

1S/C15H21F3N2O2.C4H4O4/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18;5-3(6)1-2-4(7)8/h5-8H,2-4,9-11,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b20-14+;2-1-

InChI key

LFMYNZPAVPMEGP-PIDGMYBPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluvoxamine maleate has been used as a test compound to determine the solubility and effective blood-brain barrier permeability.(2)

Biochem/physiol Actions

Fluvoxamine maleate is a selective neuronal serotonin reuptake inhibitor. It functions as an antidepressant and anti-obsessive agent. It is useful in treating obsessive compulsive disorder and panic disorder.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Solvay. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosby's Drug Reference for Health Professions null
Md Shenuarin Bhuiyan et al.
Journal of pharmacological sciences, 121(3), 177-184 (2013-02-23)
Depression is associated with a substantial increase in the risk of developing heart failure and is independently associated with increased cardiovascular morbidity and mortality. Inversely, cardiovascular disease can lead to severe depression. Thus, therapy with selective serotonin reuptake inhibitors (SSRIs)
Anton J L M van Balkom et al.
Psychotherapy and psychosomatics, 81(6), 366-374 (2012-09-12)
To compare the effectiveness of second-step treatment with cognitive therapy (CT) versus fluvoxamine in patients with obsessive-compulsive disorder (OCD) who are nonresponsive to exposure in vivo with response prevention (ERP). A 12-week randomized controlled trial at an outpatient clinic in
Eiko Sunami et al.
Internal medicine (Tokyo, Japan), 51(10), 1187-1193 (2012-06-13)
Antidepressants have been recommended for the treatment of post-stroke depression (PSD). The purpose of this study was to evaluate the effect of fluvoxamine maleate, a selective serotonin re-uptake inhibitor (SSRI), on depressive state, sleep disturbance, and serum melatonin levels in
Yumi Sugimoto et al.
European journal of pharmacology, 696(1-3), 96-100 (2012-10-09)
We studied the involvement of the sigma(1) receptor in the antidepressant-like effects of the selective serotonin reuptake inhibitor (SSRI) fluvoxamine in DBA/2 mice using the forced swimming test. The effects of the selective sigma(1) receptor antagonist N-[2-(3,4-dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino) ethylamine (BD1047) at

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service