Skip to Content
Merck

Skip To

F2379

Fetuin from fetal bovine serum

lyophilized powder, suitable for cell culture

Synonym(s):

Fetuin from fetal calf serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-762-7
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Biological source:
bovine serum (fetal)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine serum (fetal)

Quality Segment

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.3% free N-acetylneuraminic acid

solubility

H2O: 1 mg/mL, colorless to faint yellow or tan

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... FETUA(504615)

General description

Fetuin from fetal bovine serum is a 64 kDa plasma glycoprotein. It is produced by the liver and adipose tissue.

Application

Fetuin is a protein with the ability to bind to the insulin receptor and inhibit insulin action. Fetuin has been used in studies of type 2 diabetes mellitus (T2DM). In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients. Fetuin has also been used in studies to demonstrate its protective role in the development of arterial calcification.
Fetuin from fetal bovine serum has been used:
  • to synthesize primary and secondary calciprotein particles (CPP)
  • to study the binding of Xerocomus chrysenteron lectin (XCL) to the T antigen expressed by membrane proteins
  • as a supplement in Ham′s F10 media for myoblast culture.

Biochem/physiol Actions

Fetuin, an inhibitor of insulin receptor tyrosine kinase, modulates bone remodeling and calcium metabolism. It is involved in keratinocytes migration and breast tumor cell proliferative signaling. Fetuin also regulates vascular calcification, insulin resistance and protease activity It acts as a biomarker for neurodegenerative diseases. Fetuin has anti-inflammatory property.

Preparation Note

Prepared by ammonium sulfate fractionation of fetal bovine serum by the method of Pederson, K.O., J. Phys. and Colloid Chem., 51, 164 (1947).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3004F3385341506
technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, MALDI-MS: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

biological source

bovine serum (fetal)

biological source

bovine serum (fetal)

biological source

-

biological source

bovine serum

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized solid

impurities

≤0.3% free N-acetylneuraminic acid

impurities

~0.2% free N-acetylneuraminic acid

impurities

≤0.3% free N-acetylneuraminic acid

impurities

-

solubility

H2O: 1 mg/mL, colorless to faint yellow or tan

solubility

-

solubility

H2O: 1 mg/mL

solubility

water: 50 mg/mL

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Fetuin from fetal bovine serum


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. Is there information available on the content/concentration of bovine IgG and BSA in the fetuin products F2379 and F3385 (CAS 9014-81-7)?

    1 answer
    1. Testing for bovine IgG and BSA is not conducted in the final product. A purity agarose gel is performed on F2379, but quantification, testing, or reporting for IgG and BSA is not done. Additionally, the marker size used for QC testing is uncertain, and both IgG and BSA are larger than fetuin.

      Helpful?

  2. How pure is this fetuin? There's a protein content % but could there be other proteins from serum present when using ammonium sulfate fractionation?

    1 answer
    1. Product F2379 can not be considered pure Fetuin. Neither the purity, nor the identity of potential contaminating proteins, is determined.

      Helpful?

  3. Is there a good antibody to recognize this bovine fetuin B by western blot?

    1 answer
    1. Unfortunately, the Fetuin antibodies currently available are not tested against bovine. While fetuin is fairly conserved from bovine to human, reactivity cannot be predicted. See the links below for fetuin antibodies against human that have been testing for use in western blot.

      ABN1469
      https://www.sigmaaldrich.com/product/mm/abn1469

      SAB5701234
      https://www.sigmaaldrich.com/product/sigma/sab5701234

      Helpful?

Reviews

No rating value

Active Filters