Merck
F1911
Nalgene® membrane forceps
stainless steel
Synonym(s):
tongs, tweezers, Nalgene DS0399-0001, handling tool, laboratory tweezers, sample manipulator, sample pinchers
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
eCl@ss:
32035504
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Nalgene DS0399-0001
General description
The unique cross-action design holds the membrane without applying pressure that may cause damage.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)