Skip to Content
Merck

Skip To

F1506

Sodium fumarate dibasic

≥99%

Synonym(s):

Disodium fumarate, Fumaric acid disodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NaOOCCH=CHCOONa
CAS Number:
Molecular Weight:
160.04
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
241-087-7
Beilstein/REAXYS Number:
4301302
MDL number:
Assay:
≥99%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

functional group

ketone

SMILES string

[Na+].[Na+].[O-]C(=O)\C=C\C([O-])=O

InChI

1S/C4H4O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H,(H,5,6)(H,7,8);;/q;2*+1/p-2/b2-1+;;

InChI key

MSJMDZAOKORVFC-SEPHDYHBSA-L

Application

  • Enhancement of drug absorption: Sodium fumarate dibasic is utilized to improve the dissolution behavior and oral absorption of drugs with pH-dependent solubility, enhancing bioavailability and therapeutic efficacy in pharmaceutical applications (Salehi et al., 2021).
  • Development of novel drug carriers: The compound is used in the synthesis and characterization of novel injectable drug carriers, aiming to enhance the delivery and efficacy of therapeutic agents in medical treatment strategies (Guo et al., 2005).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
128422489468O0136
description

≥99%

description

95%

description

99 atom % 13C

description

BioXtra

assay

≥99%

assay

95%

assay

-

assay

≥99.0% (titration)

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

functional group

ketone

functional group

carboxylic acid, ester

functional group

-

functional group

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters