Skip to Content
Merck
All Photos(1)

Key Documents

F1395

Sigma-Aldrich

β-L-Fucose 1-phosphate bis(cyclohexylammonium) salt

≥98%

Synonym(s):

6-Deoxy-β-L-galactose 1-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O8P · 2C6H14N
Molecular Weight:
442.48
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98%

form

powder

technique(s)

thin layer chromatography (TLC): suitable

solubility

H2O: soluble 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.C[C@@H]3O[C@H](OP(O)(O)=O)[C@@H](O)[C@H](O)[C@@H]3O

InChI

1S/2C6H13N.C6H13O8P/c2*7-6-4-2-1-3-5-6;1-2-3(7)4(8)5(9)6(13-2)14-15(10,11)12/h2*6H,1-5,7H2;2-9H,1H3,(H2,10,11,12)/t;;2-,3+,4+,5-,6+/m..0/s1

InChI key

FQMPFZHILABVMA-PEJHDPODSA-N

Application

β-L-Fucose 1-phosphate is suitable as both substrate and product to identify, differentiate and characterize GTP fucose pyrophosphorylase(s) (GFPP; fucose-1-phosphate guanylyltransferases) involved in the formation of the nucleotide-sugar GDP-beta-l-fucose and other fucosylation donor substrates such as 3,3′-Diaminobenzidine (GDP)-β-l-fucose. β-L-Fucose 1-phosphate may be used to generate new fucosylation donor substrates for use in glycan fucosylation research.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonie Engels et al.
Glycobiology, 24(2), 170-178 (2013-11-20)
Fucosyltransferases (FucTs) are essential tools for the synthesis of fucosylated glycoconjugates. Multistep enzyme catalysis of fucosylated glycans is not limited as long as isolated and well-characterized FucTs are available. The present paper introduces a novel bacterial α1,2-FucT of the glycosyltransferase
Toshihisa Kotake et al.
The Journal of biological chemistry, 283(13), 8125-8135 (2008-01-18)
Monomeric sugars generated during the metabolism of polysaccharides, glycoproteins, and glycolipids are imported to the cytoplasm and converted to respective nucleotide sugars via monosaccharide 1-phosphates, to be reutilized as activated sugars. Because L-fucose (L-Fuc) is activated mainly in the form
GDP-L-fucose pyrophosphorylase. Purification, cDNA cloning, and properties of the enzyme.
Pastuszak I, Ketchum C, Hermanson G, et al.
The Journal of Biological Chemistry, 4273, 30165-30174 (1998)
Bing Ma et al.
Glycobiology, 16(12), 158R-184R (2006-09-16)
Fucosylated carbohydrate structures are involved in a variety of biological and pathological processes in eukaryotic organisms including tissue development, angiogenesis, fertilization, cell adhesion, inflammation, and tumor metastasis. In contrast, fucosylation appears less common in prokaryotic organisms and has been suggested
Stephen Quirk et al.
Biochemistry, 44(32), 10854-10863 (2005-08-10)
GTP-l-fucose pyrophosphorylase (GFPP, E. C. 2.7.7.30) catalyzes the reversible condensation of guanosine triphosphate and beta-l-fucose-1-phosphate to form the nucleotide-sugar GDP-beta-l-fucose. The enzyme functions primarily in the mammalian liver and kidney to salvage free l-fucose during the breakdown of glycolipids and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service