Skip to Content
Merck
All Photos(2)

Key Documents

F1180

Sigma-Aldrich

Anti-Flotillin 1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FLOT1, Anti-Reggie-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using human HEK-293K cells
western blot: 0.5-1 μg/mL using whole extract of mouse lung
western blot: 1-2 μg/mL using whole extract of rat lung

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FLOT1(10211)
mouse ... Flot1(14251)
rat ... Flot1(64665)

Related Categories

General description

Flotillin 1(FLOT1), also known as reggie-2, is an estrogen responsive gene mapped to human chromosome 6p21.33. This gene codes for an integral membrane protein and a component of lipid rafts.
Flotillins are lipid raft associated proteins expressed mainly in adipose tissue, striated muscle tissues, and lung. It has a crucial role in several cellular processes like phagocytosis, endocytosis, insulin signalling and neuronal regeneration. Anti-flotillin 1 antibody can be used in immunobloting and immunolocalization. Rabbit anti- flotillin 1 antibody reacts specifically with flotillin 1 of rat, mouse and human.

Immunogen

synthetic peptide corresponding to amino acid resides 413-428 of mouse flotillin 1 with an N-terminal added cysteine, conjugated to KLH. The corresponding sequence is identical in rat and differs by one amino acid in human.

Application

Anti-flotillin 1 antibody can be used in immunofluorescence and western blotting.
Anti-flotillin 1 antibody produced in rabbit has been used in immunoblot analysis.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

Flotillin 1 (FLOT1) has a role in endocytosis. It also functions in growth factor signaling and activates mitogen-activated protein kinase signaling. In addition, it is also involved in phagocytosis, regulation of actin cytoskeleton and membrane trafficking. The protein has been associated with tumor progression and is expressed in many cancers, including breast cancer.

Target description

Flotillin 1 encodes a caveolae-associated, integral membrane protein. Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett Nixon et al.
Journal of cellular physiology, 226(10), 2651-2665 (2011-07-28)
Mammalian spermatozoa attain the ability to fertilize an oocyte as they negotiate the female reproductive tract. This acquisition of functional competence is preceded by an intricate cascade of biochemical and functional changes collectively known as "capacitation." Among the universal correlates
Marlene Spitzel et al.
Cells, 11(11) (2022-06-11)
Fabry disease (FD) is a rare life-threatening disorder caused by deficiency of the alpha-galactosidase A (GLA) enzyme with a characteristic pain phenotype. Impaired GLA production or function leads to the accumulation of the cell membrane compound globotriaosylceramide (Gb3) in the
Aubin Moutal et al.
bioRxiv : the preprint server for biology (2020-09-02)
Global spread of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) continues unabated. Binding of SARS-CoV-2's Spike protein to host angiotensin converting enzyme 2 triggers viral entry, but other proteins may participate, including neuropilin-1 receptor (NRP-1). As both Spike protein and
Anastasia Chillà et al.
Molecular & cellular proteomics : MCP, 12(7), 1926-1938 (2013-04-11)
Endothelial cell caveolar-rafts are considered functional platforms that recruit several pro-angiogenic molecules to realize an efficient angiogenic program. Here we studied the differential caveolar-raft protein composition of endothelial colony-forming cells following stimulation with VEGF, which localizes in caveolae on interaction
Alberto Camacho et al.
The European journal of neuroscience, 38(5), 2672-2683 (2013-06-15)
Peroxisome proliferator-activated receptor gamma-coactivator-1 alpha (PGC1a) is involved in energy and lipid metabolism, and its loss leads to neurodegenerative changes in the striatum. Here we performed lipidomic analysis on brain extracts from PGC1a mutant and wild-type mice. We found increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service