Skip to Content
Merck

F0518

Ferrous sulfate chelate solution

100 ×, suitable for plant cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.72
UNSPSC Code:
10171502
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

Quality Level

form

solution

concentration

100 ×

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

Ferrous:EDTA chelate is a redox active form of bound iron. Ferrous:EDTA chelate will give up its iron to plant iron chelators. This iron chelate may be useful in a wide range of studies on iron metabolism and regulation in plants and plant oxidative processes.

Other Notes

Contains 2.785 g/L FeSO4 ⋅ 7H2O and 3.725g/L EDTA 2Na ⋅ 2H2O


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



T Franza et al.
Molecular plant-microbe interactions : MPMI, 12(2), 119-128 (1999-02-02)
Low iron availability is a triggering signal for coordinated expression of the genes encoding pectate lyases PelB, PelC, PelD, and PelE, and chrysobactin iron transport functions, which are two main determinants of phytopathogenicity of the Erwinia chrysanthemi strain 3937. The
Abhishek Jain et al.
Applied microbiology and biotechnology, 102(10), 4563-4575 (2018-04-05)
Nickel (Ni(II)) toxicity is addressed by many different bacteria, but bacterial responses to nickel stress are still unclear. Therefore, we studied the effect of Ni(II) toxicity on cell proliferation of α-proteobacterium Caulobacter crescentus. Next, we showed the mechanism that allows
Neeraja Vajrala et al.
BMC microbiology, 11, 37-37 (2011-02-23)
In response to environmental iron concentrations, many bacteria coordinately regulate transcription of genes involved in iron acquisition via the ferric uptake regulation (Fur) system. The genome of Nitrosomonas europaea, an ammonia-oxidizing bacterium, carries three genes (NE0616, NE0730 and NE1722) encoding



Global Trade Item Number

SKUGTIN
F0518-1L04061833611272
F0518-1LSTAB04065272810807