Skip to Content
Merck

Skip To

F0500

Fast Blue RR Salt

crystalline

Synonym(s):

4-Benzoylamino-2,5-dimethoxybenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 24

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C15H14ClN3O3 · 1/2 ZnCl2
CAS Number:
Molecular Weight:
387.89
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
238-777-5
MDL number:
Colour Index Number:
37155
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fast Blue RR Salt, crystalline

form

crystalline

Quality Segment

technique(s)

microbe id | staining: suitable

solubility

water: 1 mg/mL, clear to hazy, yellow-green

ε (extinction coefficient)

≥15000 at 326-332  nm, ≥7000 at 390-396 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc([N+]#N)c(OC)cc1NC(=O)c2ccccc2.COc3cc([N+]#N)c(OC)cc3NC(=O)c4ccccc4

InChI

1S/2C15H13N3O3.4ClH.Zn/c2*1-20-13-9-12(18-16)14(21-2)8-11(13)17-15(19)10-6-4-3-5-7-10;;;;;/h2*3-9H,1-2H3;4*1H;/q;;;;;;+2/p-2

InChI key

LLARVKAOQLUCQI-UHFFFAOYSA-L

Application

Fast blue RR salt has been used in the immunohistochemistry to detect the activity of alkaline phosphatase. It has been used in the procedure of connective tissue staining using picrosirius red.

Biochem/physiol Actions

Fast blue RR is mainly used in histological stainings. It binds to the reaction product of alkaline phosphatase and precipitates it as an insoluble compound. The sites of alkaline phosphatase activity appears black. Fast blue RR is also used for the detection of esterase activity. It couples with naphthol (released from naphthol acetate) and forms an insoluble azo-dye which appears red/blue.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F3378F2256FBS25
technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

-

technique(s)

-

form

crystalline

form

powder

form

powder

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

hematology
histology

solubility

water: 1 mg/mL, clear to hazy, yellow-green

solubility

H2O: 10 mg/mL

solubility

water: 1 mg/mL

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

500

storage temp.

−20°C

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Fast Blue RR Salt


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Culling CFA
Handbook of Histopathological and Histochemical Techniques (2013)
Mesodermal expression of Fgfr2S252W is necessary and sufficient to induce craniosynostosis in a mouse model of Apert syndrome.
Holmes G and Basilico C
Developmental Biology, 368, 283-283 (2012)
Philip F Dobson et al.
Scientific reports, 10(1), 11643-11643 (2020-07-17)
The pathogenesis of declining bone mineral density, a universal feature of ageing, is not fully understood. Somatic mitochondrial DNA (mtDNA) mutations accumulate with age in human tissues and mounting evidence suggests that they may be integral to the ageing process.



Global Trade Item Number

SKUGTIN
F0500-25G04061835347636

Questions

Reviews

No rating value

Active Filters