Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.44
UNSPSC Code:
12352203
Conjugate:
FITC conjugate
Clone:
PY20, monoclonal
Application:
EIA, IF (d), IP, WB
Citations:
1
biological source
mouse
Quality Segment
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
PY20, monoclonal
form
buffered aqueous glycerol solution
technique(s)
direct immunofluorescence: 5-10 μg/mL, enzyme immunoassay: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG2b
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Immunogen
phosphotyrosine-protein conjugate
Physical form
Solution in 20 mM sodium phosphate and 150 mM sodium chloride, pH 7.5, containing 50% glycerol and 3 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F0426-100UG | 04061833384718 |