Key Documents
EP5810757002
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 4 x 18-30 mL tubes, set of 2 pieces
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
for use with 4 x 18-30 mL tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810757002
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL rectangular buckets
General description
Tube adapter for Eppendorf 250 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG