Skip to Content
Merck
All Photos(1)

Key Documents

EMU077471

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Thbd

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCCAGGCTCTTACTCCTGTATGTGTGAGACAGGCTACCAGTTGGCTGCAGACGGACACCGGTGTGAGGACGTGGATGACTGTAAGCAGGGGCCCAATCCATGTCCCCAGCTCTGTGTTAACACCAAGGGCGGCTTCGAATGCTTCTGCTATGATGGCTATGAGTTGGTGGATGGAGAGTGCGTGGAGCTTCTGGATCCGTGTTTCGGATCTAACTGCGAGTTTCAGTGCCAGCCAGTGAGCCCCACCGACTACCGATGCATCTGCGCTCCAGGCTTCGCACCCAAGCCGGATGAACCGCACAAGTGCGAAATGTTCTGCAATGAAACTTCGTGCCCAGCAGACTGTGACCCTAACTCTCCTACTGTTTGTGAATGCCCTGAAGGCTTCATCCTGGACGAGGGTTCCGTATGCACGGACATTGATGAGTGCAGTCAAGGCGAATGCTTCAC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minttu Kansikas et al.
Human mutation, 35(9), 1123-1127 (2014-06-14)
Lynch syndrome (LS), the most common familial colon cancer, is associated with mismatch repair (MMR) malfunction. As mutation carriers inherit one normal and one defected MMR gene allele, cancer risk can be considered as limited amount of normal MMR gene
Tadashi Inoue et al.
Human molecular genetics, 23(21), 5672-5682 (2014-06-09)
Latent TGF-β-binding protein-2 (LTBP-2) is an extracellular matrix protein associated with microfibrils. Homozygous mutations in LTBP2 have been found in humans with genetic eye diseases such as congenital glaucoma and microspherophakia, indicating a critical role of the protein in eye
O Richmond et al.
Veterinary microbiology, 180(3-4), 223-229 (2015-10-09)
Porcine circovirus type 2 (PCV2) and porcine reproductive and respiratory syndrome virus (PRRSV) continue to have a negative economic impact on global swine production operations. Host immune modulations that potentiate disease during PCV2 and/or PRRSV infections are important areas of
Chun-Te Wu et al.
BMC cancer, 14, 375-375 (2014-06-03)
The identification of potential tumor markers will help improve therapeutic planning and patient management. Thrombomodulin (TM) is a sensitive urothelial marker. TM was reported to be one of the endogenous anti-metastatic factors and has diagnostic and prognostic values for the
Devanshi Seth et al.
World journal of gastroenterology, 20(36), 13088-13104 (2014-10-04)
To investigate over-expression of Osteopontin (OPN) pathway expression and mechanisms of action in human alcoholic liver disease (ALD), in vivo and in vitro acute alcohol models. OPN pathway was evaluated in livers from patients with progressive stages of human ALD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service