Merck
Skip To
EL1
Endura-Luers
Stopcocks for vacuum manifold
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NA.52
UNSPSC Code:
41105501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Quality Segment
General description
Sigma offers Endura-Luer stopcocks to be used in conjunction with the VM20 Vacuum Manifold. As an alternative to plugs, they create maximum efficiency when purifying multiple preps. Endura-Luers resist chemical erosion, cracking, and breaking.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Questions
  1. Of what material is this product made? Polypropylene? Is there any PTFE or PFAS? Is it suitable for use in PFAS testing?
    1 answer
    1. These Kynar stopcocks are not recommended for PFAS testing.
      Helpful?