Skip to Content
Merck
All Photos(1)

Key Documents

EHU156431

Sigma-Aldrich

MISSION® esiRNA

targeting human TIMP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGTTGTTGCTGTGGCTGATAGCCCCCAGCAGGGCCTGCACCTGTGTCCCACCCCACCCACAGACGGCCTTCTGCAATTCCGACCTCGTCATCAGGGCCAAGTTCGTGGGGACACCAGAAGTCAACCAGACCACCTTATACCAGCGTTATGAGATCAAGATGACCAAGATGTATAAAGGGTTCCAAGCCTTAGGGGATGCCGCTGACATCCGGTTCGTCTACACCCCCGCCATGGAGAGTGTCTGCGGATACTTCCACAGGTCCCACAACCGCAGCGAGGAGTTTCTCATTGCTGGAAAACTGCAGGATGGACTCTTGCACATCACTACCTGCAGTTTTGTGGCTCCCTGGAACAGCCTGAGCTTAGCTCAGCGCCGGGGCTTCACCAAGACCTACACTGTTGGCTGTGAGGAATGCACAGTGTTTCCCTGTTTATCCATCCCCTGCAAAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu-Mei Huang et al.
Journal of dermatological science, 96(3), 159-167 (2019-11-26)
Macrophages play important roles during wound healing, and delayed healing in diabetics is associated with sustained inflammation. M1 type macrophage is recognized to secrete excessive amount of tumor necrosis factor-alpha (TNF-α) as compared to its M2 counterpart. We hypothesized that
Omar M Omar et al.
Molecular carcinogenesis, 57(11), 1577-1587 (2018-07-24)
Tissue inhibitor matrix metalloproteinase-1 (TIMP1) is one of four identified members of the TIMP family. We evaluated the role of TIMP1 in gastric cancer using human and mouse tissues along with gastric organoids and in vitro cell models. Using quantitative
Fenglin Zhao et al.
International journal of molecular medicine, 44(5), 1609-1618 (2019-09-06)
Parastomal hernia (PH) is a common complication following stoma formation. Abnormal collagen synthesis has been suggested to be involved in PH. The aim of the present study is to explore the effect and mechanism of the collagen synthesis on PH.
Rosemarie Chirco D'Angelo et al.
Molecular cancer research : MCR, 12(9), 1324-1333 (2014-06-05)
Tissue inhibitor of metalloproteinase-1 (TIMP-1) regulates intracellular signaling networks for inhibition of apoptosis. Tetraspanin (CD63), a cell surface binding partner for TIMP-1, was previously shown to regulate integrin-mediated survival pathways in the human breast epithelial cell line MCF10A. In the
Jingshu Tang et al.
Acta pharmaceutica Sinica. B, 10(6), 987-1003 (2020-07-10)
Blood-brain barrier (BBB) breakdown and the associated microvascular hyperpermeability are hallmark features of several neurological disorders, including traumatic brain injury (TBI). However, there is no viable therapeutic strategy to rescue BBB function. Tissue inhibitor of metalloproteinase-1 (TIMP1) has been considered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service