Skip to Content
Merck
All Photos(1)

Key Documents

EHU139921

Sigma-Aldrich

MISSION® esiRNA

targeting human KAT5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTCACCCGGATGAAGAACATTGAGTGCATTGAGCTGGGCCGGCACCGCCTCAAGCCGTGGTACTTCTCCCCGTACCCACAGGAACTCACCACATTGCCTGTCCTCTACCTGTGCGAGTTCTGCCTCAAGTACGGCCGTAGTCTCAAGTGTCTTCAGCGTCATTTGACCAAGTGTGACCTACGACATCCTCCAGGCAATGAGATTTACCGCAAGGGCACCATCTCCTTCTTTGAGATTGATGGACGTAAGAACAAGAGTTATTCCCAGAACCTGTGTCTTTTGGCCAAGTGTTTCCTTGACCATAAGACACTGTACTATGACACAGACCCTTTCCTCTTCTACGTCATGACAGAGTATGACTGTAAGGGCTTCCACATCGTGGGCTACTTCTCCAAGGAGAAAGAATCAACGGAAGACTACAATGTGGCCTGCATCCTAAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Li et al.
PloS one, 6(8), e23725-e23725 (2011-08-23)
GPR50 is an orphan G-protein coupled receptor most closely related to the melatonin receptors. The physiological function of GPR50 remains unclear, although our previous studies implicate the receptor in energy homeostasis. Here, we reveal a role for GPR50 as a
Xueqin Wang et al.
BMC microbiology, 10, 228-228 (2010-08-28)
Salmonella enterica is a facultative intracellular pathogen that replicates within a membrane-bound compartment termed Salmonella containing vacuole (SCV). The biogenesis of SCV requires Salmonella type III protein secretion/translocation system and their effector proteins which are translocated into host cells to
Zhaoxiang Yu et al.
Archives of biochemistry and biophysics, 693, 108550-108550 (2020-08-30)
Bromodomain-containing 8 (BRD8), which belongs to the histone acetyl transferase (HAT) complex, functions as a driver in colorectal cancer. However, the role of BRD8 and its related regulatory mechanisms in hepatocellular carcinoma (HCC) remain unexplored. In this study, we found
Shengyuan Zeng et al.
Protein & cell, 8(3), 202-218 (2016-10-16)
UHRF2 is a ubiquitin-protein ligase E3 that regulates cell cycle, genomic stability and epigenetics. We conducted a co-immunoprecipitation assay and found that TIP60 and HDAC1 interact with UHRF2. We previously demonstrated that UHRF2 regulated H3K9ac and H3K14ac differentially in normal
Mathieu Dalvai et al.
PLoS genetics, 9(4), e1003387-e1003387 (2013-05-03)
Histone variants, including histone H2A.Z, are incorporated into specific genomic sites and participate in transcription regulation. The role of H2A.Z at these sites remains poorly characterized. Our study investigates changes in the chromatin environment at the Cyclin D1 gene (CCND1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service