Skip to Content
Merck
All Photos(1)

Key Documents

EHU112661

Sigma-Aldrich

MISSION® esiRNA

targeting human BNIP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGACGGAGTAGCTCCAAGAGCTCTCACTGTGACAGCCCACCTCGCTCGCAGACACCACAAGATACCAACAGAGCTTCTGAAACAGATACCCATAGCATTGGAGAGAAAAACAGCTCACAGTCTGAGGAAGATGATATTGAAAGAAGGAAAGAAGTTGAAAGCATCTTGAAGAAAAACTCAGATTGGATATGGGATTGGTCAAGTCGGCCGGAAAATATTCCCCCCAAGGAGTTCCTCTTTAAACACCCGAAGCGCACGGCCACCCTCAGCATGAGGAACACGAGCGTCATGAAGAAAGGGGGCATATTCTCTGCAGAATTTCTGAAAGTTTTCCTTCCATCTCTGCTGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianjie Li et al.
International journal of molecular medicine, 46(2), 729-739 (2020-07-07)
Long non‑coding RNA (lncRNA) DGCR5 has been identified as a tumor suppressor in several types of cancer. However, its biological functions in pancreatic cancer (PaCa) have not yet been fully elucidated. The present study was designed to investigate the role
Zhiliang He et al.
Acta biochimica et biophysica Sinica, 49(1), 25-32 (2016-11-20)
Nutrition deficiency is reported to induce apoptosis of chondrocytes and degeneration of cartilage endplate (CEP) in rabbit. Cartilage endplate stem cells (CESCs) are important for the integrity of structure and function of CEP. Bcl-2/adenovirus E1B 19-kDa-interacting protein 3 (BNIP3) has
Zong-Jie Fu et al.
Redox biology, 36, 101671-101671 (2020-08-24)
In the present study, we hypothesized that hypoxia-inducible factor 1α (HIF-1α)-mediated mitophagy plays a protective role in ischemia/reperfusion (I/R)-induced acute kidney injury (AKI). Mitophagy was evaluated by measuring the changes of mitophagy flux, mitochondria DNA copy number, and the changes
Chongcheng Wang et al.
Cell death & disease, 11(8), 630-630 (2020-08-18)
Induction of lethal autophagy has become a strategy to eliminate glioma cells, but it remains elusive whether autophagy contributes to cell death via causing mitochondria damage and nuclear translocation of apoptosis inducing factor (AIF). In this study, we find that
Jie Liu et al.
Molecular medicine reports, 16(5), 7253-7260 (2017-09-26)
Nutrient deprivation (ND)‑induced nucleus pulposus (NP) cell death serves an important role in intervertebral disc degeneration disease. However, the underlying mechanisms have yet to be thoroughly elucidated. The present study created a cell culture model under ND conditions to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service