Skip to Content
Merck
All Photos(1)

Key Documents

EHU081481

Sigma-Aldrich

MISSION® esiRNA

targeting human MITF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGGGGTTTATTTCAGCAAACTTGTTGAATTTATTTTTAAGAAAGAAATACTGTATTGGGAAGTTACTGTTACTTGATAACAATGTTTTAACAAGAAGCAATGTTATAAAGTTAGTTTCAGTGCATTATCTACTTGTGTAGTCCTATGCAATAACAGTAGTGTTACATGTATCAAGCCTAGATGTTTTATACAGATGCCATATAGTGTTATGAGCCAGGCTGTTGAATGGAATTTCTCAGTAGCAGCCTACAACTGAATAGCAAGTGGCATAAAGCATATCCATTCAGAATGAAGTGCCTTAAATATAGCAGTAGTCTTTTTTGGACTAGCACTGACTGAACTGTAATGTAGGGGAAAGTTTCATGATGGTATCTATAGTCAAGACGAACATGTAGCATGGTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megha Rajasekhar et al.
Scientific reports, 8(1), 7264-7264 (2018-05-10)
Myelopoiesis involves differentiation of hematopoietic stem cells to cellular populations that are restricted in their self-renewal capacity, beginning with the common myeloid progenitor (CMP) and leading to mature cells including monocytes and granulocytes. This complex process is regulated by various
Hua Chen et al.
International journal of clinical and experimental pathology, 13(4), 730-737 (2020-05-02)
Coronary atherosclerosis affects human health all over the world. PON1 was found to be associated with coronary atherosclerosis but the specific mechanism is still unclear. Non-coding RNA plays an important role in many diseases. In recent years, studies have focused
Jing Wang et al.
Journal of cellular biochemistry, 119(11), 8862-8871 (2018-08-21)
Oral lichen planus (OLP) is a severe T cell-mediated disorder of the mucosa, which causes chronic inflammation. Forkhead box P3 (Foxp3) regulates the immune response and plays an important role in immunological diseases. The current study aimed to determine the
Paola Falletta et al.
Genes & development, 31(1), 18-33 (2017-01-18)
The intratumor microenvironment generates phenotypically distinct but interconvertible malignant cell subpopulations that fuel metastatic spread and therapeutic resistance. Whether different microenvironmental cues impose invasive or therapy-resistant phenotypes via a common mechanism is unknown. In melanoma, low expression of the lineage
Yue Chang et al.
Cancer cell international, 21(1), 29-29 (2021-01-09)
Endometrial cancer is an invasive gynecological cancer prevalent in the world. The pathogenesis of endometrial cancer is related to multiple levels of regulation, referring to oestrogen, tumor-suppressor gene (e.g. PTEN) or microRNAs (e.g. miR-23a and miR-29b). Metapristone is a hormone-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service