Skip to Content
Merck
All Photos(1)

Key Documents

EHU064501

Sigma-Aldrich

MISSION® esiRNA

targeting human CD82

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATGGTCCTGTCCATCTGCTTGTGCCGGCACGTCCATTCCGAAGACTACAGCAAGGTCCCCAAGTACTGAGGCAGCTGCTATCCCCATCTCCCTGCCTGGCCCCCAACCTCAGGGCTCCCAGGGGTCTCCCTGGCTCCCTCCTCCAGGCCTGCCTCCCACTTCACTGCGAAGACCCTCTTGCCCATCCTGACTGAAAGTAGGGGGCTTTCTGGGGCCTAGCGATCTCTCCTGGCCTATCCGCTGCCAGCCTTGAGCCCTGGCTGTTCTGTGGTTCCTCTGCTCACCGCCCATCAGGGTTCTCTTAGCAACTCAGAGAAAAATGCTCCCCACAGCGTCCCTGGCGCAGGTGGGCTGGACTTCTACCTGCCCTCAAGGGTGTGTATATTGTATAGGGGCAACTGTATGAAAAATTGGGGAGGAGGGGGCCGGGCGCGGTGGCTCACGCCTGTAATCCCAGCACTTTGGGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianwen Long et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 102, 1195-1202 (2018-05-02)
Melanoma has been a severe threat to human health, microRNAs play vital roles in the oncogenesis and progression of cancers. In this report, the roles and mechanism of miR-338-5p were investigated in the development of melanoma. A total of 46
Muskan Floren et al.
Oncogene, 39(19), 3910-3925 (2020-03-24)
A principal challenge in treating acute myeloid leukemia (AML) is chemotherapy refractory disease. As such, there remains a critical need to identify key regulators of chemotherapy resistance in AML. In this study, we demonstrate that the membrane scaffold, CD82, contributes
Chao Huang et al.
Journal of extracellular vesicles, 9(1), 1692417-1692417 (2019-12-07)
Tumour metastasis suppressor KAI1/CD82 inhibits tumour cell movement. As a transmembrane protein, tetraspanin CD82 bridges the interactions between membrane microdomains of lipid rafts and tetraspanin-enriched microdomains (TEMs). In this study, we found that CD82 and other tetraspanins contain cholesterol recognition/interaction
Qing-Hui Zhang et al.
Digestive diseases and sciences, 60(7), 1967-1976 (2015-02-06)
This study was to investigate the effects and mechanisms of miR-362-3p on regulation of gastric cancer (GC) cell metastasis potential. We detected miR-362-3p level in GC and adjacent normal tissues and investigated the relationship with clinicopathological factors. Next, we analyzed
Thomas B Layton et al.
Nature communications, 11(1), 2768-2768 (2020-06-04)
Fibrotic disorders are some of the most devastating and poorly treated conditions in developed nations, yet effective therapeutics are not identified for many of them. A major barrier for the identification of targets and successful clinical translation is a limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service