Skip to Content
Merck
All Photos(1)

Key Documents

EHU062171

Sigma-Aldrich

MISSION® esiRNA

targeting human IRF4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAGCCAAGCATAAGGTCTGCCGAAGCCTTGGCGTTCTCAGACTGCCGGCTGCACATCTGCCTGTACTACCGGGAAATCCTCGTGAAGGAGCTGACCACGTCCAGCCCCGAGGGCTGCCGGATCTCCCATGGACATACGTATGACGCCAGCAACCTGGACCAGGTCCTGTTCCCCTACCCAGAGGACAATGGCCAGAGGAAAAACATTGAGAAGCTGCTGAGCCACCTGGAGAGGGGCGTGGTCCTCTGGATGGCCCCCGACGGGCTCTATGCGAAAAGACTGTGCCAGAGCAGGATCTACTGGGACGGGCCCCTGGCGCTGTGCAACGACCGGCCCAACAAACTGGAGAGAGACCAGACCTGCAAGCTCTTTGACACACAGCAGTTCTTGTCAGAGCTGCAAGCGTTTGCTCACCACGGCCGCTCCCTGCCAAGATTCCAGGTGACTCTATGCTTTGGAGAGGAGTTTCCAGACCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Watanabe et al.
Mucosal immunology, 7(6), 1312-1325 (2014-03-29)
It is well established that polymorphisms of the caspase activation and recruitment domain 15 (CARD15) gene, a major risk factor in Crohn's disease (CD), lead to loss of nucleotide-binding oligomerization domain 2 (NOD2) function. However, a molecular explanation of how
Sorim Nam et al.
Journal of leukocyte biology, 100(6), 1273-1284 (2016-09-08)
Myeloid-derived suppressor cells (MDSCs) are immature cells that do not differentiate into mature myeloid cells. Two major populations of PMN-MDSCs (Ly6G
Takuya Yashiro et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(10), 11481-11491 (2019-07-18)
C-C chemokine receptor type 7 (CCR7) is essential for migration of dendritic cells (DCs) to draining lymph nodes. PU.1/Spi1 is a transcription factor playing a critical role in the gene regulation of DCs. PU.1 knockdown decreased the expression of CCR7
Diego Barriales et al.
PLoS biology, 19(1), e3001062-e3001062 (2021-01-05)
Lyme carditis is an extracutaneous manifestation of Lyme disease characterized by episodes of atrioventricular block of varying degrees and additional, less reported cardiomyopathies. The molecular changes associated with the response to Borrelia burgdorferi over the course of infection are poorly
Takuya Yashiro et al.
Scientific reports, 9(1), 1161-1161 (2019-02-06)
The chemokine CCL22 is predominantly produced by dendritic cells (DCs) and macrophages. CCL22 acts on CCR4-expressing cells including Th2 and Treg. Although a correlation between the CCL22-CCR4 axis and allergic diseases has been established, the mechanism of monocyte lineage-specific Ccl22

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service