Select a Size
About This Item
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AACGTGCACCTTTCTCATCCTGTTTATGTCCGACGTGCAGCTACTGAAAATATTCCTGTGGTTAGAAGACCTGATCGAAAAGATCTACTTGGATATCTCAATGGTGAAGCGTCAACATCGGCAAGTATAGACAGAAGCGCTCCCTTAGAAATAGGTCTTCAGCGATCTACTCAAGTCAAACGAGCTGCAGATGAAGTTTTAGCAGAAGCAAAGAAACCACGAATTGAGGATGAAGAGTGTGTGCGCCTTGATAAAGAGAGATTGGCTGCCCGTTTGGAGGGTCACAAAGAAGGGATTGTACAGACTGAACAGATTAGGTCTTTGTCTGAAGCTATGTCAGTGGAAAAAATTGCTGCAATCAAAGCCAAAATTATGGCTAAGAAAAGATCTACTATCAAGACTGATCTAGATGATGACATAACTGCCCTTAAACAGAGGAGTTTTGTGGATGCTGAGGTAGATGTGACCCG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... CDC73(79577)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.