Skip to Content
Merck
All Photos(1)

Key Documents

EHU030271

Sigma-Aldrich

MISSION® esiRNA

targeting human MECP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGTGACCGAGAGAGTTAGCTGACTTTACACGGAGCGGATTGCAAAGCAAACCAACAAGAATAAAGGCAGCTGTTGTCTCTTCTCCTTATGGGTAGGGCTCTGACAAAGCTTCCCGATTAACTGAAATAAAAAATATTTTTTTTTCTTTCAGTAAACTTAGAGTTTCGTGGCTTCAGGGTGGGAGTAGTTGGAGCATTGGGGATGTTTTTCTTACCGACAAGCACAGTCAGGTTGAAGACCTAACCAGGGCCAGAAGTAGCTTTGCACTTTTCTAAACTAGGCTCCTTCAACAAGGCTTGCTGCAGATACTACTGACCAGACAAGCTGTTGACCAGGCACCTCCCCTCCCGCCCAAACCTTTCCCCCATGTGGTCGTTAGAGACAGAGCGACAGAGCAGTTGAGAGGACACTCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuta Tanimoto et al.
Endocrinology, 156(1), 58-70 (2014-11-05)
The growth factor progranulin is as an important regulator of transformation in several cellular systems. We have previously demonstrated that progranulin acts as an autocrine growth factor and stimulates motility, proliferation, and anchorage-independent growth of castration-resistant prostate cancer cells, supporting
V O Okoh et al.
British journal of cancer, 112(10), 1687-1702 (2015-05-13)
17β-Oestradiol (E2)-induced reactive oxygen species (ROS) have been implicated in regulating the growth of breast cancer cells. However, the underlying mechanism of this is not clear. Here we show how ROS through a novel redox signalling pathway involving nuclear respiratory
Anja Rogler et al.
Journal of cancer research and clinical oncology, 141(10), 1779-1790 (2015-03-04)
We previously showed that the Wnt-signaling antagonist SFRP1 (secreted frizzled-related protein 1) is a promising marker in bladder cancer. The aim of this study was to validate the prognostic role and analyze the functional significance of SFRP1. Four bladder cancer
Balaram Thota et al.
Journal of neurosurgery, 121(2), 374-383 (2014-06-01)
Insulin-like growth factor binding proteins (IGFBPs) have been implicated in the pathogenesis of glioma. In a previous study the authors demonstrated that IGFBP-3 is a novel glioblastoma biomarker associated with poor survival. Since signal transducer and activator of transcription 1
Riddhi Atul Jani et al.
Journal of cell science, 128(17), 3263-3276 (2015-07-26)
Melanosomes are a class of lysosome-related organelles produced by melanocytes. Biogenesis of melanosomes requires the transport of melanin-synthesizing enzymes from tubular recycling endosomes to maturing melanosomes. The SNARE proteins involved in these transport or fusion steps have been poorly studied.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service