Skip to Content
Merck
All Photos(1)

Key Documents

EHU020361

Sigma-Aldrich

MISSION® esiRNA

targeting human CUL4A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCTTGTGTGGAGAAACAGCTATTAGGAGAACATTTAACAGCAATTCTGCAGAAAGGGCTCGACCACTTACTGGATGAGAACAGAGTGCCGGACCTCGCACAGATGTACCAGCTGTTCAGCCGGGTGAGGGGCGGGCAGCAGGCGCTGCTGCAGCACTGGAGCGAGTACATCAAGACTTTTGGAACAGCGATCGTAATCAATCCTGAGAAAGACAAAGACATGGTCCAAGACCTGTTGGACTTCAAGGACAAGGTGGACCACGTGATCGAGGTCTGCTTCCAGAAGAATGAGCGGTTCGTCAACCTGATGAAGGAGTCCTTTGAGACGTTCATCAACAAGAGACCCAACAAGCCTGCAGAACTGATCGCAAAGCATGTGGATTCAAAGTTAAGAGCAGGCAACAAAGAAGCCACAGACGAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yidan Ren et al.
Respiratory research, 20(1), 84-84 (2019-05-08)
Chronic obstructive pulmonary disease (COPD) is a common respiratory disease with high morbidity and mortality. The most important pathophysiological change of COPD is airway obstruction. Airway obstruction can cause airflow restriction and obstructive ventilation dysfunction. Currently, many studies have shown
Hiroshi Nakade et al.
International journal of clinical oncology, 25(3), 446-455 (2019-09-20)
Cullin4A (CUL4A), which is a component of E3 ubiquitin ligase, is implicated in many cellular events. Although the altered expression of CUL4A has been reported in several human cancers, the role of CUL4A in esophageal cancer remains unknown. We investigated
B Englinger et al.
British journal of cancer, 116(4), 489-500 (2017-01-18)
Colorectal carcinoma (CRC) is the third most common cancer worldwide. Platinum-based anticancer compounds still constitute one mainstay of systemic CRC treatment despite limitations due to adverse effects and resistance development. Trabectedin has shown promising antitumor effects in CRC, however, again
Laura P Saucedo-Cuevas et al.
Oncotarget, 5(8), 2330-2343 (2014-05-30)
The CUL4A E3 ubiquitin ligase is involved in the regulation of many cellular processes and its amplification and/or overexpression has been observed in breast cancer. The 13q34 amplification, which is associated with the basal-like breast cancer subtype, has been proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service