Skip to Content
Merck
All Photos(1)

Documents

EHU019931

Sigma-Aldrich

MISSION® esiRNA

targeting human KIF11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCCGTAACAAGAGAGGAGTGATAATTAAAGGTTTAGAAGAAATTACAGTACACAACAAGGATGAAGTCTATCAAATTTTAGAAAAGGGGGCAGCAAAAAGGACAACTGCAGCTACTCTGATGAATGCATACTCTAGTCGTTCCCACTCAGTTTTCTCTGTTACAATACATATGAAAGAAACTACGATTGATGGAGAAGAGCTTGTTAAAATCGGAAAGTTGAACTTGGTTGATCTTGCAGGAAGTGAAAACATTGGCCGTTCTGGAGCTGTTGATAAGAGAGCTCGGGAAGCTGGAAATATAAATCAATCCCTGTTGACTTTGGGAAGGGTCATTACTGCCCTTGTAGAAAGAACACCTCATGTTCCTTATCGAGAATCTAAACTAACTAGAATCCTCCAGGATTCTCTTGGAGGGCGTACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
Advanced healthcare materials, 5(12), 1493-1504 (2016-04-26)
Developing RNA-interference-based therapeutic approaches with efficient and targeted cytosolic delivery of small interfering RNA (siRNA) is remaining a critical challenge since two decades. Herein, a multifunctional transferrin receptor (TfR)-targeted siRNA delivery system (Tf&INF7) is designed based on siRNA complexes formed
Kai Zhu et al.
American journal of translational research, 11(4), 2245-2256 (2019-05-21)
Micro RNA (miRNAs) is a kind of non coding small RNAs with negative regulation function, which plays an important role in regulating the occurrence and development of tumors. In this study, we analyzed the expression level and role of miRNA-186-5p
Takeharu Imai et al.
Anticancer research, 37(1), 47-55 (2016-12-25)
Oesophageal squamous cell carcinoma (ESCC) and colorectal cancer (CRC) are common types of human cancer. Spheroid colony formation is used to characterize cancer stem cell (CSCs). In the present study, we analyzed the significance of kinesin family 11 (KIF11 in
Myles Fennell et al.
Assay and drug development technologies, 13(7), 347-355 (2015-08-13)
Uptake of nutrients, such as glucose and amino acids, is critical to support cell growth and is typically mediated by cell surface transporters. An alternative mechanism for the bulk uptake of nutrients from the extracellular space is macropinocytosis, a nonclathrin
Benjamin Steinborn et al.
The journal of gene medicine, 20(7-8), e3041-e3041 (2018-06-28)
Developing new drug delivery carriers addressing chemoresistance is still full of challenges and opportunities. As the rapid development of small interfering RNA (siRNA) provides promising therapeutic perspectives, nanocarriers for drug and siRNA co-delivery present new alternatives for cancer therapy. A

Articles

esiRNA are endoribonuclease prepared siRNAs that target the same mRNA sequence for gene silencing. Here are some of the most asked questions regarding esiRNA uses and availability.

esiRNA are endoribonuclease prepared siRNAs that target the same mRNA sequence for gene silencing. Here are some of the most asked questions regarding esiRNA uses and availability.

esiRNA are endoribonuclease prepared siRNAs that target the same mRNA sequence for gene silencing. Here are some of the most asked questions regarding esiRNA uses and availability.

esiRNA are endoribonuclease prepared siRNAs that target the same mRNA sequence for gene silencing. Here are some of the most asked questions regarding esiRNA uses and availability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service