Skip to Content
Merck

ECM001

Human Fibronectin

recombinant, expressed in HEK 293 cells, powder, suitable for cell culture

Synonym(s):

Fibronectin, CIG, Cold-insoluble globulin, FN

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75
MDL number:
Biological source:
human
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Mol wt:
259.5 kDa (The protein migrates as a higher band on SDS-PAGE due to glycosylation.)
Assay:
≥95% (SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fibronectin human, recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in HEK 293 cells

sterility

sterile

assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

259.5 kDa (The protein migrates as a higher band on SDS-PAGE due to glycosylation.)

packaging

pkg of 200 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg endotoxin, tested

solubility

water: soluble

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... FN1(2335)

General description

Fibronectin (FN) is a glycoprotein with 5-9% carbohydrates present as N- and O-linked oligosaccharides. It belongs to the ligand glycoprotein family. The FN gene is localized to human chromosome 2q35. FN exists as two major isoforms, plasma-based soluble and extracellular matrix (ECM)-based insoluble form. Fibronectin structure comprises repeats of FNI, FNII and FNIII repeats. It also has fibrin-binding sites and exists as a dimer. Alternative splicing of the FN1 gene leads to the generation of many isoforms. This fibronectin protein is manufactured in human cells using an all-human production system with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports better stability of the protein in culture.

Application

Fibronectin human has been used for coating cell culture plates for culturing endothelial progenitor cells. It has also been used as a component of Dulbecco′s modified Eagle′s medium (DMEM) in the in vitro matrigel invasion assay of glioblastoma cells.
Epithelial cells, mesenchymal cells, neuronal cells, fibroblasts, neural crest cells, endothelial cells.
Recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/ml. Optimal concentration depends on cell type as well as the application or research objectives.

Biochem/physiol Actions

Fibronectin (FN) is essential for embryonic development and mediates cell adhesion and migration functions. It mediates apoptosis in nasopharyngeal carcinoma (NPC). The plasma FN regulates thrombosis. Fibronectin functions as a ligand for the integrin receptor family and contains collagen, glycoprotein and glycosaminoglycan binding sites. Mutation in the FNI gene leads to an imbalance in the soluble and insoluble forms of fibronectin leading to glomerular deposits in hereditary kidney disease, glomerulopathy. Missense mutations in the FN1 gene are implicated in the skeletal dysplasias disorder, Spondylometaphyseal dysplasias.

Features and Benefits

  • Human-derived fibronectin
  • Produced through recombinant expression in HEK 293 cells
  • Low endotoxin levels
  • Absence of bacterial contamination

Physical form

Supplied as a powder, lyophilized from CAPS buffered saline.

Analysis Note

The biological activity of recombinant human fibronectin was tested in culture by measuring the ability of immobilized fibronectin to support adhesion of CHO cells.


Still not finding the right product?

Explore all of our products under Fibronectin human


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library