Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C9H20N2O4S
CAS Number:
Molecular Weight:
252.33
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
240-198-8
Beilstein/REAXYS Number:
3957385
MDL number:
Quality Level
assay
≥99.5% (titration)
form
powder
pH
7.3-8.7
useful pH range
7.3-8.7
pKa (25 °C)
8.0
pKa
8
mp
237-239 °C (lit.)
solubility
H2O: 25 g + 50 mL, clear, colorless
λmax
A290 at ≤0.1 (C=33% w/w)
application(s)
diagnostic assay manufacturing
SMILES string
OCCN1CCN(CCCS(O)(=O)=O)CC1
InChI
1S/C9H20N2O4S/c12-8-7-11-5-3-10(4-6-11)2-1-9-16(13,14)15/h12H,1-9H2,(H,13,14,15)
InChI key
OWXMKDGYPWMGEB-UHFFFAOYSA-N
Application
EPPS has been used:
- to verify the effect of pH variation on the proteolytic activity of the posterior midgut (PMG) contents and anterior midgut (AMG) tissue homogenate
- to prepare a destaining buffer to destain intensely stained gel pieces
- to prepare digestion buffer for in-gel enzymatic digestion
- to reconstitute the digested peptides for the processing of brain-derived extracellular vesicle (EV) proteins and 16-plex isobaric tandem mass tag (TMT) peptide labeling
Still not finding the right product?
Explore all of our products under EPPS
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Bernhard Sissolak et al.
Journal of industrial microbiology & biotechnology, 46(8), 1167-1178 (2019-06-09)
Recombinant monoclonal antibodies are predominantly produced in mammalian cell culture bioprocesses. Post-translational modifications affect the micro-heterogeneity of the product and thereby influence important quality attributes, such as stability, solubility, pharmacodynamics and pharmacokinetics. The analysis of the surface charge distribution of
YoungSoo Kim et al.
Science advances, 5(4), eaav1388-eaav1388 (2019-04-20)
Detection of amyloid-β (Aβ) aggregates contributes to the diagnosis of Alzheimer disease (AD). Plasma Aβ is deemed a less invasive and more accessible hallmark of AD, as Aβ can penetrate blood-brain barriers. However, correlations between biofluidic Aβ concentrations and AD
Bianca Santos Henriques et al.
Frontiers in physiology, 11, 509310-509310 (2021-02-02)
Rhodnius prolixus is one important vector for the parasite Trypanosoma cruzi in Latin America, where Chagas disease is a significant health issue. Although R. prolixus is a model for investigations of vector-parasite interaction and transmission, not much has been done
Global Trade Item Number
| SKU | GTIN |
|---|---|
| E9502-100G | 04061833608999 |
| E9502-1KG | 04061833609002 |
| E9502-10G | 04061832547992 |
| E9502-250G | 04061833609019 |
| E9502-5KG | 04061833609033 |
| E9502-25G | 04061833609026 |