Skip to Content
Merck

E9145

Estrone 3-sulfate potassium salt

≥98%

Synonym(s):

1,3,5(10)-Estratrien-17-one 3-sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H21O5SK
CAS Number:
Molecular Weight:
388.52
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
214-985-1
MDL number:
Form:
powder
Assay:
≥98%
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98%

form

powder

contains

~35% Tris as stabilizer

color

off-white to light tan

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

[K].[H][C@]12CC[C@]3(C)C(=O)CC[C@@]3([H])[C@]1([H])CCc4cc(OS(O)(=O)=O)ccc24

InChI

1S/C18H22O5S.K.H/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;;/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);;/t14-,15-,16+,18+;;/m1../s1

InChI key

XFQXTMKDDYWSBS-DLEHHZMWSA-N

Biochem/physiol Actions

The most abundant estrogen in adult human circulation. Conversion to estradiol requires steroid sulfatase activity. This activity was used to evaluate freshly resected bone fragments against cultured osteoblasts and osteosearcoma cells as models for studying human bone metabolism.

Packaging

Package size based on steroid content


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Maria Karlgren et al.
Journal of medicinal chemistry, 55(10), 4740-4763 (2012-05-01)
The hepatic organic anion transporting polypeptides (OATPs) influence the pharmacokinetics of several drug classes and are involved in many clinical drug-drug interactions. Predicting potential interactions with OATPs is, therefore, of value. Here, we developed in vitro and in silico models
L M M C Sousa et al.
Placenta, 33(10), 788-794 (2012-07-31)
Our objectives were to investigate the possible role of VEGFA in bovine placenta steroid synthesis and to determine whether cloned derived placental cells present similar responses as non-cloned ones. Placental cells from cloned (term) and non-cloned (days 90, 150, 210
Anna Koenen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 47(4), 774-780 (2012-09-18)
Previously, the steroid hormone progesterone has been demonstrated to stimulate OATP2B1-mediated transport of estrone-3-sulphate (E(1)S), dehydroepiandrosterone sulphate (DHEAS) and pregnenolone sulphate (PS), which may influence the uptake of precursor molecules for steroid hormone synthesis. However, it is unclear whether OATP2B1



Global Trade Item Number

SKUGTIN
E9145-100MG04061833608937