Skip to Content
Merck
All Photos(1)

Documents

E8280

Sigma-Aldrich

Exo 1

≥98% (HPLC), solid

Synonym(s):

2-(4-Fluorobenzoylamino)benzoic acid methyl ester, 2-(4-Fluorobenzoylamino)methylbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12NFO3
CAS Number:
Molecular Weight:
273.26
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 19 mg/mL

SMILES string

COC(=O)c1ccccc1NC(=O)c2ccc(F)cc2

InChI

1S/C15H12FNO3/c1-20-15(19)12-4-2-3-5-13(12)17-14(18)10-6-8-11(16)9-7-10/h2-9H,1H3,(H,17,18)

InChI key

KIAPWMKFHIKQOZ-UHFFFAOYSA-N

Application

Exo 1 has been used as a component of the BRB80 buffer to study coat protein 1 (COPI)-mediated membrane trafficking. It has also been used as a Golgi inhibitor to study its effects on the long interspersed nuclear element-1 (L1) retrotransposition.

Biochem/physiol Actions

Exo 1 is a cell-permeable methylanthranilate analog that promotes the release of adenosine diphosphate (ADP)-ribosylation factor (ARF) 1 from Golgi membranes. It also inhibits the membrane trafficking between the endoplasmic reticulum (ER) and the Golgi bodies.
Reversible inhibitor of exocytosis; Golgi ARF 1 (ADP-Ribosylation Factor) GTPase activator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Feng et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(11), 6469-6474 (2003-05-10)
A phenotypic screen was used to search for drug-like molecules that can interfere with specific steps in membrane traffic. 2-(4-Fluorobenzoylamino)-benzoic acid methyl ester (Exo1), identified in this screen, induces a rapid collapse of the Golgi to the endoplasmic reticulum, thus
ALan F Rees et al.
Marine biology, 164(2), 30-30 (2017-01-31)
Many marine megavertebrate taxa, including sea turtles, disperse widely from their hatching or birthing locations but display natal homing as adults. We used flipper tagging, satellite tracking and genetics to identify the origin of loggerhead turtles living in Amvrakikos Gulf
Justin Chun et al.
Molecular biology of the cell, 19(8), 3488-3500 (2008-06-06)
Despite extensive work on ADP-ribosylation factor (Arf) 1 at the Golgi complex, the functions of Arf2-5 in the secretory pathway, or for that of any Arf at the ER-Golgi intermediate compartment (ERGIC) remain uncharacterized. Here, we examined the recruitment of
Time-lapse observation of chromosomes, cytoskeletons and cell organelles during male meiotic divisions in Drosophila
Tanabe K, et al.
Bio-protocol, 7, e2225-e2225 (2017)
Philip J Lester et al.
Scientific reports, 10(1), 12398-12398 (2020-07-28)
CRISPR gene drives have potential for widespread and cost-efficient pest control, but are highly controversial. We examined a potential gene drive targeting spermatogenesis to control the invasive common wasp (Vespula vulgaris) in New Zealand. Vespula wasps are haplodiploid. Their life

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service