Skip to Content
Merck

E8029

E-Toxate Endotoxin standard

for endotoxin quantitation

Synonym(s):

E. coli 0.55:B5 LPS standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

contains

solubilizer

storage temp.

2-8°C

Application

E-Toxate endotoxin standard was used along with the limulus amebocyte lysate assay kit to determine the concentration of endotoxins in isolated samples of human holotransferrin and apotransferrin used to treat primary hepatocyte cultures.

Analysis Note

Standardized against USP reference standard endotoxin (RSE)

Other Notes

A preparation of E. coli 055:B5 lipopolysaccharide
Can be stored at 2-8 °C when stored separately from the E-Toxate kit.

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Akrivi Chrysanthopoulou et al.
Clinical immunology (Orlando, Fla.), 247, 109240-109240 (2023-01-25)
Kruppel-like factor 2 (KLF2) has been linked with fibrosis and neutrophil-associated thromboinflammation; however, its role in COVID-19 remains elusive. We investigated the effect of disease microenvironment on the fibrotic potential of human lung fibroblasts (LFs) and its association with KLF2
Anna Scanu et al.
International journal of molecular sciences, 23(21) (2022-11-12)
We investigated the effects of bactericidal/permeability-increasing protein (BPI) alone or in combination with hyaluronic acid (HA) in two animal models: collagen-induced arthritis (CIA) and crystal-induced inflammation. In CIA, mice were intraperitoneally injected with PBS, HA, or BPI plus or minus
Lan Lin et al.
Blood, 110(6), 2182-2189 (2007-06-02)
The peptide hormone hepcidin is the principal regulator of systemic iron homeostasis. We examined the pathway by which iron stimulates the production of hepcidin. In humans who ingested 65 mg of iron, the increase in transferrin saturation preceded by hours



Global Trade Item Number

SKUGTIN
E8029-1VL04061838163301