Skip to Content
Merck

Skip To

E4643

Heparin-Binding EGF-Like Growth Factor human

≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

hHB-EGF, HB-EGF, Heparin-Binding Epidermal Growth Factor-Like Growth Factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf21 cells
Assay:
≥97% (SDS-PAGE and N-terminal analysis)
Form:
lyophilized powder
Mol wt:
9.5 kDa
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Heparin-Binding EGF-Like Growth Factor human, HB-EGF, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

assay

≥97% (SDS-PAGE and N-terminal analysis)

form

lyophilized powder

potency

≤3 ng/mL

quality

endotoxin tested

mol wt

9.5 kDa

packaging

pkg of 50 and 250 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Biochem/physiol Actions

Heparin-binding epidermal growth factor (HB-EGF), like BTC, is a heparin-binding EGF family member. Natural soluble HB-EGF is a 19 kDa glycoprotein of multiple forms due to heterogeneous truncations. Soluble HB-EGF is mitogenic and chemotactic for fibroblasts, keratinocytes, and smooth muscle cells but not capillary endothelial cells. Production of HB-EGF is detected in monocytes, macrophages, and vascular and aortic smooth muscle cells (SMC). HB-EGF may play a role in the pathogenesis of atherosclerosis. It may also be an effector of Jun-induced oncogenic transformation. HB-EGF binds to EGFR with high affinity and to ErbB with moderate affinity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in the EGF-responsive BALB/3T3 cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E9644H9661P5208
biological source

human

biological source

human

biological source

human

biological source

human

assay

≥97% (SDS-PAGE and N-terminal analysis)

assay

≥97% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥97% (SDS-PAGE)

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in E. coli

recombinant

expressed in NSO cells

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters