Skip to Content
Merck

Skip To

E2888

Earle′s Balanced Salts

With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EBSS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: 0.011 g/L
glucose: 1.0 g/L (Dextro)
NaHCO3: 2.2 g/L

shipped in

ambient

storage temp.

room temp

General description

Balanced Salt solution participates in providing a buffer system to maintain the medium in optimum pH range (7.2-7.6), irrigating and diluting while maintaining the intra- and extracellular osmotic balance. It helps to maintain the normal cell metabolism by giving water and bulk inorganic ions. Balanced Salt solution provides the energy source for cell metabolism by supplying carbohydrates such as glucose.

Application

Earle′s Balanced Salts has been used:
  • to incubate hepatocellular carcinoma cell line[1]
  • as a component of embryo culture medium for washing rabbit embryos[2]
  • to culture mouse embryonic neuroretina[3]


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E6267E3024E7510
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

liquid

form

liquid

form

liquid

form

liquid

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

components

phenol red: 0.011 g/L
glucose: 1.0 g/L (Dextro)
NaHCO3: 2.2 g/L

components

NaHCO3: 2.2 g/L
phenol red: 0.011 g/L
glucose: 1.0 g/L (Dextro)

components

phenol red: no
NaHCO3: 2.2 g/L
glucose: 1.0 g/L (Dextro)

components

glucose: 10.0 g/L
NaHCO3: no
phenol red: 0.11 g/L

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


Still not finding the right product?

Explore all of our products under Earle′s Balanced Salts


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters