Skip to Content
Merck

E1411

Proline Specific Endopeptidase from Flavobacterium sp.

lyophilized powder, ≥5.0 units/mg solid

Synonym(s):

EndoProC, Endoprolylpeptidase, Endoproteinase Pro-C, PEPase, Prolyl Endopeptidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥5.0 units/mg solid
Biological source:
bacterial (Flavobacterium spp.)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Flavobacterium spp.)

form

lyophilized powder

specific activity

≥5.0 units/mg solid

mol wt

~78 kDa

shipped in

dry ice

storage temp.

−20°C

Application

Proline specific endopeptidase is useful for the determination of amino acid sequences of peptides and proteins containing proline residues. Proline specific endopeptidases may be useful in cancer, neurology and diabetes research . Prolyl endopeptidase (PEP) may be used as a drug target for neuropsychiatric diseases such as stress disorder, depression, and schizophrenia .

Biochem/physiol Actions

An endopeptidase is a proteolytic peptidase that breaks peptide bonds of nonterminal amino acids. Product E1411 is specific for proline (Pro). The Prolyl endopeptidase (PEP) mechanism is thought to be an induced fit mechanism, where the native enzyme exists in a conformationally flexible opened state but is changed to a closed state upon substrate binding .

Physical form

White amorphous lyophilized powder

Other Notes

One unit causes the formation of one micromole of p-nitroaniline per minute under specific pH and temperature conditions.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library