Skip to Content

E1253

Estriol

synthetic (organic), ≥97%, immunomodulatory , powder

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol, 16α-Hydroxyestradiol, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
200-022-2
MDL number:
Beilstein/REAXYS Number:
2508172
Assay:
≥97%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Estriol, ≥97%

biological source

synthetic (organic)

Quality Segment

sterility

non-sterile

assay

≥97%

form

powder

mp

280-282 °C (lit.)

solubility

pyridine: 50 mg/mL, clear, colorless to faintly brownish-yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1

InChI key

PROQIPRRNZUXQM-ZXXIGWHRSA-N

Gene Information

human ... SERPINA6(866)
mouse ... Esr1(13982)
rat ... Ar(24208)

Application

Estriol has been used as a sex hormone to determine its binding constants to insulin (INS), the insulin receptor (IR), and INS-like peptides derived from the IR.

Biochem/physiol Actions

Estriol is a metabolite of 17β-estradiol with much lower biological activity. It is the primary estrogen found in the urine. Large quantities of estriol and estrone are produced by the placenta during pregnancy. These are also the primary estrogens produced by adipose tissue in men and post-menopausal women. Estriol may exhibit immunomodulatory benefits against various diseases such as autoimmune, inflammatory, and neurodegenerative conditions.


Still not finding the right product?

Explore all of our products under Estriol


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library