Skip to Content
Merck

DUO96030

Duolink® PLA Multicolor Probemaker Kit - Orange

Proximity Ligation Assay Multiplexing: protein-protein interaction, post-translational modification and low abundant protein events

Synonym(s):

in situ Proximity Ligation Assay Kit, Protein Protein Interaction Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41105331
Fluorescence:
λex 542 nm; λem 562 nm (orange), λex 542 nm; λem 562 nm
Technique(s):
immunofluorescence: suitable, multiplexing: suitable, proximity ligation assay: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Duolink®

technique(s)

immunofluorescence: suitable, multiplexing: suitable, proximity ligation assay: suitable

fluorescence

λex 542 nm; λem 562 nm (orange), λex 542 nm; λem 562 nm

suitability

suitable for fluorescence

shipped in

dry ice

storage temp.

−20°C

Application

Specificity
Orange fluorescence detection reagents are often used with Cy®3 filter.

Application Note
Primary antibodies are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC), or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

Multicolor Duolink® proximity ligation assay (PLA®) is an application of the Duolink® technology that allows for endogenous, simultaneous detection of up to 4 protein interactions, post-translational modifications, or protein expression levels (including low abundant protein) in fixed cells and tissue samples. Follow the Duolink® PLA Probemaker Guide for Multicolor Detection and the Duolink® PLA Multicolor Detection protocol to use this product.

  • Visualize and detect multiple proteins events in one experiment.
  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Detection of low abundant proteins due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Faster and more simple than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Components
Duolink® PLA Multicolor Probemaker Kit - Orange contains all the necessary reagents to perform the conjugations of Orange Oligo F and Orange Oligo G directly to primary antibodies. The resulting PLA probes (Orange F and Orange G) must be used together and cannot be paired with any other Duolink® PLA probe.

This product is comprised of the following:
  • Orange Oligo F DUO86030F
  • Orange Oligo G DUO86030G
  • Conjugation Buffer DUO82033
  • Stop Reagent DUO82034
  • Storage Solution DUO82035

See datasheet for more information.

To perform a Multicolor Duolink® PLA experiment primary antibodies (PLA, IHC, ICC or IF validated) that recognize protein targets of interest are required. Duolink® PLA Multicolor Probemaker-generated probes (Red A/Red B, Green C/Green D, Orange F/Orange G, and/or FarRed H/FarRed I) are used in combination to perform the Multiplex experiment.The reagents in the Duolink® PLA Multicolor Reagent Pack are required to perform the amplification and detection of the resulting PLA probes. Required additional reagents include Duolink® Wash Buffers and Mounting Medium.

Legal Information

Cy is a registered trademark of Cytiva
Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library