Skip to Content
Merck

D9164

1,2-Dilinoleoyl-3-oleoyl-rac-glycerol

≥98%

Synonym(s):

1,2-Di(cis-9,12-octadecadienoyl)-3-(cis-9-octadecenoyl)-rac-glycerol, OLL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C57H100O6
CAS Number:
Molecular Weight:
881.40
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥98%

form

liquid

functional group

ester, ketone

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(OC(COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)CCCCCCC/C=C\C/C=C\CCCCC

InChI

1S/C57H100O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16,18-19,21,25-30,54H,4-15,17,20,22-24,31-53H2,1-3H3/b19-16-,21-18-,28-25-,29-26-,30-27-

InChI key

VVEBTVMJPTZDHO-WECKWCTPSA-N

Application


  • Research on the effect of spleen-invigorating and anti-swelling active ingredients in crude and processed coix seed based on Spectrum - Effects relationship combined with chemometrics.: This study examines the active ingredients in crude and processed coix seed and their effects on spleen invigoration and anti-swelling. The analysis was performed using a combination of chemometrics and spectrum-effect relationships, providing insights into the biochemical interactions and potential therapeutic applications of 1,2-Dilinoleoyl-3-oleoyl-rac-glycerol in pharmaceutical formulations. (Du et al., 2021).



Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library