Skip to Content
Merck
All Photos(1)

Documents

D8428

Sigma-Aldrich

Decorin from bovine articular cartilage

salt-free, lyophilized powder

Synonym(s):

Decorin protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine articular cartilage

Quality Level

form

salt-free, lyophilized powder

mol wt

~100 kDa

packaging

glass bottle of 0.5 mg

technique(s)

cell culture | mammalian: suitable

solubility

PBS: ≥2.00 mg/mL, clear, colorless to yellow

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... DCN(280760)

General description

Decoran is a small proteoglycan composed of a chondroitin/dermatan sulfate glycosaminoglycans attached to a 40 kDa core protein. It is found primarily in the extracellular matrix of articular cartilage. Decorin interacts with collagen types I and II via its core protein wherein it delays fibrillogenesis. Decorin also interacts with fibronectin, thrombospondin and TGF-β isoforms, and the EGF-receptor.
Decorin from bovine articular cartilage is a proteoglycan that belongs to the small leucine-rich repeat proteoglycans and proteins (SLRPs) family. Decorin is present in almost all tissues. It has a protein core composed of leucine-rich repeats (LRRs), flanked by two cysteine-rich regions.

Application

Decorin from bovine articular cartilage has been used in solid phase binding assay (ELISA), binding assays, binding inhibition assay, parasite binding assays, scaffold fabrication, quartz crystal microbalance assay, inhibition of CSA (chondroitin sulphate A) binding assay and glycans and glycan biotinylation.

Biochem/physiol Actions

Decorin plays an important role in guiding the proper assembly of collagen during fibrillogenesis. Deficiency in decorin results in altered tissue structure and function. It controls growth factors like EGF and TGF-β. Decorin blocks aggregation of fibrils into fibers and fibrillogenesis in cell-free constructs of polymerized type I collagen.

Other Notes

Decorin is an approx. 100 kDa proteoglycan consisting of a 40 kDa core protein and one chondroitin or dermatan sulfate glycosaminoglycan chain. From 0.2 μm filtered solution prior to lyophilization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chondroitin sulphate A-binding site of the VAR2CSA protein involves multiple N-terminal domains.
Dahlback M, et al.
The Journal of Biological Chemistry (2011)
Xiaolei Wang et al.
Scientific reports, 7(1), 9672-9672 (2017-08-31)
Choroidal neovascularization(CNV) is the most severe complication in Age-related macular degeneration(AMD) and the most common cause of irreversible blindness in the elderly in developed world. The aim of this study was to identify the effect of transforming growth factor-β(TGF-β) and
Seda Duran G Ler et al.
Indian journal of ophthalmology, 69(2), 369-373 (2021-01-20)
The aims of this study were to evaluate the effects of decorin (DCN) in rat oxygen-induced retinopathy (OIR) model and to compare the results with those of bevacizumab. Twenty-eight newborn Sprague-Dawley rats were randomly divided into four groups. Group I
Structural and functional insight into how the Plasmodium falciparum VAR2CSA protein mediates binding to chondroitin sulfate A in placental malaria.
Clausen T M, et al.
The Journal of Biological Chemistry, 287(28), 23332-23345 (2012)
Caroline Pehrson et al.
Vaccine, 35(4), 610-618 (2016-12-26)
Malaria in pregnancy is associated with significant morbidity in pregnant women and their offspring. Plasmodium falciparum infected erythrocytes (IE) express VAR2CSA that mediates binding to chondroitin sulphate A (CSA) in the placenta. Two VAR2CSA-based vaccines for placental malaria are in

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service