Skip to Content
Merck
All Photos(5)

Key Documents

D8417

Sigma-Aldrich

4′,6-Diamidino-2-phenylindole dihydrochloride

powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence

Synonym(s):

2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
Beilstein:
4894417
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

Assay

≥98% (HPLC and TLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 20 mg/mL
PBS: insoluble

ε (extinction coefficient)

30 at 263 nm in H2O

fluorescence

λex 340 nm; λem 488 nm (nur DAPI)
λex 364 nm; λem 454 nm (DAPI-DNA-Komplex)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cl.Cl.NC(=N)c1ccc(cc1)-c2cc3ccc(cc3[nH]2)C(N)=N

InChI

1S/C16H15N5.2ClH/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;;/h1-8,21H,(H3,17,18)(H3,19,20);2*1H

InChI key

FPNZBYLXNYPRLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4′,6-Diamidino-2-phenylindole dihydrochloride is a fluorescent dye for staining DNA. It strongly interacts with the dAT-rich (deoxy adenine-thymine) regions of the DNA.
Heat or sonication may be required. Solutions stored in the dark at room temperature or 4 °C should be stable for 2 to 3 weeks.

Application

4′,6-Diamidino-2-phenylindole dihydrochloride has been used in staining of various cells in immunocytochemistry and cytochemistry, human subcutaneous adipose tissues for fluorescence imaging, embryo sections for immunofluorescence.
DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels.
DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma.

Biochem/physiol Actions

Cell permeable fluorescent minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA (preferentially to AT rich DNA), forming a stable complex which fluoresces approximately 20 times greater than DAPI alone.

Caution

Protect from light.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endogenous Nodal signaling regulates germ cell potency during mammalian testis development
Spiller C M, et al.
Development, 139(22), 4123-4132 (2012)
Behavior of mesenchymal stem cells stained with 4', 6-diamidino-2-phenylindole dihydrochloride (DAPI) in osteogenic and non osteogenic cultures
Ocarino N M, et al.
Biocell, 32(2), 175-183 (2008)
Interaction of 4?-6-diamidino-2-phenylindole to nucleic acids, and its implication to their template activity in RNA-polymerase reaction of E. coli bacteria and of friend-virus infected mouse spleen
Mildner B, et al.
Cancer letters, 4(2), 89-98 (1978)
IFATS collection: Human adipose tissue-derived stem cells induce angiogenesis and nerve sprouting following myocardial infarction, in conjunction with potent preservation of cardiac function
Cai L, et al.
Stem Cells, 27(1), 230-237 (2009)
Flavivirus NS4A-induced autophagy protects cells against death and enhances virus replication
McLean J E, et al.
Test, 286(25), 22147-22159 (2011)

Articles

Detect mycoplasma contamination in cell culture through the PCR, DNA stain, or culture tests. Discover mycoplasma prevention, elimination, and detection kits.

Detect mycoplasma contamination in cell culture through the PCR, DNA stain, or culture tests. Discover mycoplasma prevention, elimination, and detection kits.

Detect mycoplasma contamination in cell culture through the PCR, DNA stain, or culture tests. Discover mycoplasma prevention, elimination, and detection kits.

Detect mycoplasma contamination in cell culture through the PCR, DNA stain, or culture tests. Discover mycoplasma prevention, elimination, and detection kits.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service