Skip to Content
Merck
All Photos(1)

Documents

D8399

Sigma-Aldrich

Danazol

≥98% (TLC), powder, androgen receptor agonist

Synonym(s):

17β-Hydroxy-2,4,17α-pregnadien-20-yno[2,3-d]isoxazole, 2,4,17α-Pregnadien-20-yno[2,3-d]­isoxa­zol-17-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27NO2
CAS Number:
Molecular Weight:
337.46
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

Danazol, ≥98%

Assay

≥98%

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

originator

Sanofi Aventis

SMILES string

CC12Cc3cnoc3C=C1CCC4C2CCC5(C)C4CC[C@@]5(O)C#C

InChI

1S/C22H27NO2/c1-4-22(24)10-8-18-16-6-5-15-11-19-14(13-23-25-19)12-20(15,2)17(16)7-9-21(18,22)3/h1,11,13,16-18,24H,5-10,12H2,2-3H3/t16?,17?,18?,20?,21?,22-/m0/s1

InChI key

POZRVZJJTULAOH-ZIKCMSSPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Danazol is a weak androgen; anterior pituitary suppressant.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Hughes et al.
The Cochrane database of systematic reviews, (1)(1), CD000069-CD000069 (2007-01-27)
The synthetic androgen Danazol, was developed in the 1970's as a treatment for endometriosis. Its use was soon advocated in women with unexplained subfertility. Two randomised trials were subsequently conducted to assess the effectiveness of danazol in this population. The
V Selak et al.
The Cochrane database of systematic reviews, (4)(4), CD000068-CD000068 (2007-10-19)
Endometriosis is defined as the presence of endometrial tissue (stromal and glandular) outside the normal uterine cavity. Conventional medical and surgical treatments for endometriosis aim to remove or decrease the deposits of ectopic endometrium. The observation that hyper androgenic states
Terry Gernsheimer et al.
Blood, 121(1), 38-47 (2012-11-15)
A mild thrombocytopenia is relatively frequent during pregnancy and has generally no consequences for either the mother or the fetus. Although representing no threat in the majority of patients, thrombocytopenia may result from a range of pathologic conditions requiring closer
Hywel D Williams et al.
Journal of pharmaceutical sciences, 101(9), 3360-3380 (2012-05-31)
The Lipid Formulation Classification System Consortium is an industry-academia collaboration, established to develop standardized in vitro methods for the assessment of lipid-based formulations (LBFs). In this first publication, baseline conditions for the conduct of digestion tests are suggested and a
Johannes Parmentier et al.
International journal of pharmaceutics, 437(1-2), 253-263 (2012-09-04)
Liposomes are generally well tolerated drug delivery systems with a potential use for the oral route. However, little is known about the fate of liposomes during exposure to the conditions in the gastro-intestinal tract (GIT). To gain a better understanding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service