Skip to Content
Merck

D8388

Dimethyl pimelimidate dihydrochloride

powder

Synonym(s):

DMP, Dimethyl pimelinediimidate dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3OC(=NH)CH2CH2]2CH2 · 2HCl
CAS Number:
Molecular Weight:
259.17
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
261-320-6
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

reaction suitability

reagent type: cross-linking reagent

solubility

H2O: 10 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.COC(=N)CCCCCC(=N)OC

InChI

1S/C9H18N2O2.2ClH/c1-12-8(10)6-4-3-5-7-9(11)13-2;;/h10-11H,3-7H2,1-2H3;2*1H

InChI key

LRHXBHUTQWIZTN-UHFFFAOYSA-N

General description

Dimethyl pimelimidate dihydrochloride is a homobifunctional cross-linking reagent used in crosslinking, protein interaction and protein modification. It typically reacts with primary amines in the pH range 7.0-10.0 to form amidine bonds.

Application

Dimethyl pimelimidate dihydrochloride has been used to develop a technique for isolating Enterocytozoon bieneusi spores directly from human stool samples with a high degree of purity, which opens up new approaches for studying this parasite.

Other Notes

Note that the amidine linkage preserves original primary amine positive charge. Incorporates a seven atom linker.

Disclaimer

The reagent is readily hydrolyzed at neutral pH.


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library