Skip to Content
Merck

Skip To

D8132

Dehydro-L-(+)-ascorbic acid dimer

≥80% (enzymatic)

Synonym(s):

Bis(dehydro-L-ascorbic acid), Bis-DHA, DHA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H12O12
CAS Number:
Molecular Weight:
348.22
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Beilstein/REAXYS Number:
84277
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥80% (enzymatic)

form

powder

color

faint beige to beige, faint brown to light brown, white to yellow

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@@H](C(=O)C(=O)C1=O)[C@@H](O)CO

InChI

1S/C6H6O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-8H,1H2/t2-,5+/m0/s1

InChI key

SBJKKFFYIZUCET-JLAZNSOCSA-N

General description

Dehydro-L-(+)-ascorbic acid dimer is a water soluble molecule, which is produced by vitamin C in the lumen of gastrointestinal tract.

Application

Dehydro-L-(+)-ascorbic acid dimer has been used to study its transformation and stability under the influence of pH, concentration and temperature.

Biochem/physiol Actions

Dehydro-L-(+)-ascorbic acid dimer is used to regenerate ascorbic acid(AA). AA oxidation to dehydroascorbic acid (DHA) takes place in extracellular fluid and cells. AA acts as an antioxidant and enzyme cofactor. It is used in a wide variety of applications, including cell culture. It also acts as a reducing agent, that helps reduce oxidative stress. L-Ascorbate can be regenerated from dehydroascorbic acid (DHA) by biological systems.

Other Notes

Dimer in crystalline form; hydrated monomer in aqueous solution

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
26155695212A2218
description

≥80% (enzymatic)

description

-

description

tested according to Ph. Eur.

description

meets USP testing specifications

biological source

synthetic

biological source

-

biological source

synthetic

biological source

synthetic

assay

≥80% (enzymatic)

assay

-

assay

99.0-100.5%

assay

99.0-100.5%

form

powder

form

powder

form

solid

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

color

faint beige to beige, white to yellow, faint brown to light brown

color

-

color

white to off-white

color

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters