Skip to Content
Merck

D8037

Driselase Basidiomycetes sp.

BioReagent, suitable for plant cell culture

Synonym(s):

Driselase from Basidiomycetes sp.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
10171502
EC Number:
286-055-3
NACRES:
NA.72
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Level

form

powder

composition

Protein, ≥10% biuret

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

−20°C

Application

Driselase Basidiomycetes sp. has been used:

  • in spheroplast preparation from Coccomyxa cells
  • in a CRISPR/Cas9-based mutagenesis protocol for Brachypodium distachyon and its allopolyploid relative, Brachypodium hybridum
  • for cell wall digestion to perform whole-mount immunolocalization of Lotus japonicus root tissue

Biochem/physiol Actions

Driselase is a natural mixture of enzyme activities (fungal carbohydrates) used to digest plant cell walls to facilitate the maceration of plant materials, protoplast formation, and extraction processes. Driselase releases cell wall carbohydrates. This formulation contains enzyme activities of cellulose, endo-1,3-β-glucanase, and xylanase.

Other Notes

Crude powder containing laminarinase, xylanase and cellulase.

Legal Information

Driselase is a trademark of ASKA Animal Health Co. Ltd.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Dieter Hackenberg et al.
Plant physiology, 172(2), 1154-1166 (2016-08-24)
In this study, we report the functional characterization of heterotrimeric G-proteins from a nonvascular plant, the moss Physcomitrella patens. In plants, G-proteins have been characterized from only a few angiosperms to date, where their involvement has been shown during regulation
M C Ralet et al.
Carbohydrate research, 263(2), 227-241 (1994-10-17)
Cell walls from sugar-beet pulp contain some feruloyl groups linked to the pectic neutral side-chains. Enzymic as well as chemical hydrolysis of the pulp yielded a series of feruloylated oligosaccharides, which have been purified by Sephadex LH-20 and Biogel P-2
G J McDougall et al.
Carbohydrate research, 219, 123-132 (1991-10-14)
The retention times of 10 oligosaccharides, generated from the xyloglucans of Rosa and Tropaeolum by the action of Trichoderma cellulase, and of 17 related carbohydrates, in h.p.l.c. on an amino-substituted silica (Amino-Spheri-5) depended largely on the number of hydroxyl groups



Global Trade Item Number

SKUGTIN
D8037-1G04061833589120
D8037-5G04061833589137