Skip to Content
Merck

D7816

1,2-Di-O-lauryl-rac-glycero-3-glutaric acid 6′-methylresorufin ester

lipase substrate, solid

Synonym(s):

Chromogenic lipase substrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

1,2-Di-O-lauryl-rac-glycero-3-glutaric acid 6′-methylresorufin ester, lipase substrate

Quality Segment

form

solid

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCOCC(COC(=O)CCCC(=O)Oc1ccc2N=C3C=CC(=O)C=C3Oc2c1C)OCCCCCCCCCCCC

InChI

1S/C45H69NO8/c1-4-6-8-10-12-14-16-18-20-22-31-50-34-38(51-32-23-21-19-17-15-13-11-9-7-5-2)35-52-43(48)25-24-26-44(49)53-41-30-29-40-45(36(41)3)54-42-33-37(47)27-28-39(42)46-40/h27-30,33,38H,4-26,31-32,34-35H2,1-3H3

InChI key

JEGLJLCZRAWOBI-UHFFFAOYSA-N

Application

Chromogenic substrate for lipase

Biochem/physiol Actions

1,2-Di-O-lauryl-rac-glycero-3-glutaric acid 6′-methylresorufin ester (DGGR) is more pancreas-specific. Hence the DGGR lipase assay may be preferred as a substitute to spec cPL in diagnostic cases with suspected pancreatitis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
D7816-10MG04061832923840