Skip to Content
Merck

D6694

Anti-DSCR1 (C-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ADAP78, Anti-Calcipressin 1, Anti-DSC1, Anti-Down Syndrome Candidate Region 1, Anti-MCIP1, Anti-Myocyte-enriched calcineurin-interacting protein 1, Anti-RCAN1, Anti-RCN1, Anti-Regulator of calcineurin 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.44
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
39
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~26 kDa

species reactivity

mouse, rat, human

concentration

~1.5 mg/mL

technique(s)

western blot: 2-4 μg/mL using HEK-293T cells expressing human DSCR1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RCAN1(1827)
mouse ... Dscr1(54720)
rat ... Dscr1(266766)

General description

DSCR1 (Down Syndrome Candidate Region 1 is encoded by a gene mapped to human chromosome 21q22.1. DSCR1 belongs to a family of conserved proteins named regulators of calcineurin (RCAN), that in humans include RCAN, encoded by the DSCR1L1-2 genes. DSCR1 is highly expressed in the central nervous system (CNS) during early embryonic development, in heart, and in skeletal muscle.

Immunogen

Synthetic peptide corresponding to a sequence at C-terminal of human DSCR1, conjugated to KLH. This sequence is identical in human DSCR1 isoforms a and b, and rat and mouse DSCR1.

Application

Anti-DSCR1 (C-terminal) antibody is suitable for use in western blot (2-4 μg/mL using HEK-293T cells expressing human DSCR1) and in immunoblotting (approx. 26 kDa).
Anti-DSCR1 (C-terminal) antibody produced in rabbit has been used in proximity ligation assay (PLA) and immunoblotting.

Biochem/physiol Actions

Anti-DSCR1 (C-terminal) antibody is specific for DSCR1 in rats, mice and humans. The immunizing peptide specifically inhibits the staining of DSCR1 by immunoblotting.
Down Syndrome Candidate Region 1 (DSCR1) is involved in cellular adaptation to oxidative stress, and interacts with calcineurin to inhibit its phosphatase activity, thereby regulating the calcineurin- Nuclear factor of activated T-cells (NFAT) signaling pathway and affecting central nervous system (CNS) development and angiogenesis. Chronic overexpression of DSCR1 has been implicated in additional pathological conditions including Alzheimer′s disease and cardiac hypertrophy, possibly through inhibition of calcineurin activity. DSCR1 is induced by vascular endothelial growth factor (VEGF) in activated endothelial cells and regulates the expression of inflammatory marker genes such as interleukin-8 (IL-8), tissue factor, E-selectin and cyclooxygenase (Cox-2).
Down Syndrome Candidate Region 1 or DSCR1 is a candidate gene involved in the pathogenesis of Down′s Syndrome (DS). DSCR1 is usually expressed in cardiac, skeletal and brain tissues, whereas its overexpression in fetal brain tissues is associated with DS. DSCR1 inhibits calcineurin-mediated mRNA synthesis.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

wgk

nwg

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library