Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H11Cl2N · HCl
CAS Number:
Molecular Weight:
264.58
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: ≥10 mg/mL
originator
Merck & Co., Inc., Kenilworth, NJ, U.S.
storage temp.
−20°C
SMILES string
Cl.Clc1ccc(cc1Cl)C23CNCC2C3
InChI
1S/C11H11Cl2N.ClH/c12-9-2-1-7(3-10(9)13)11-4-8(11)5-14-6-11;/h1-3,8,14H,4-6H2;1H
InChI key
KAGBHVBIOJBGBD-UHFFFAOYSA-N
Biochem/physiol Actions
Serotonin-norepinephrine-dopamine reuptake inhibitor.
Triple reuptake inhibitor (TRI); serotonin-norepinephrine-dopamine reuptake inhibitor (SNDRI); IC50 values of ~20, 14, and 78 nM, respectively.
Features and Benefits
This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
